Skip to Content
Merck
All Photos(1)

Key Documents

112860

Sigma-Aldrich

4′-Chloro-2,2,2-trifluoroacetophenone

99%

Synonym(s):

4-Chloro-α,α,α-trifluoroacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCF3
CAS Number:
Molecular Weight:
208.56
Beilstein:
1955904
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

refractive index

n20/D 1.489 (lit.)

bp

180-183 °C (lit.)

mp

24-26 °C (lit.)

density

1.398 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

FC(F)(F)C(=O)c1ccc(Cl)cc1

InChI

1S/C8H4ClF3O/c9-6-3-1-5(2-4-6)7(13)8(10,11)12/h1-4H

InChI key

DYPQUENOGZXOGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4′-Chloro-2,2,2-trifluoroacetophenone forms a 1:2 complex with alpha-cyclodextrin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service