Merck
  • Simvastatin impairs the induction of pulmonary fibrosis caused by a western style diet: a preliminary study.
Simvastatin impairs the induction of pulmonary fibrosis caused by a western style diet: a preliminary study.
Journal of cellular and molecular medicine (2015-08-26)
Peter Kruzliak, David L Hare, Vaclav Zvonicek, Jan Klimas, Anthony Zulli
ABSTRACT
The role of an atherogenic diet in causing pulmonary fibrosis has received little attention and simvastatin has been shown to reduce pulmonary fibrosis in animal models. To determine if an atherogenic diet can induce pulmonary fibrosis and whether simvastatin treatment is beneficial by up-regulating heat shock protein 70 and 90. New Zealand white rabbits (n = 15) were divided: Group 1 (control); Group 2 (MC) received a normal rabbit diet with 1% methionine plus 0.5% cholesterol (atherogenic diet). Group 3 received the same diet as the MC group plus 5 mg/kg/day simvastatin orally (MCS). After 4 weeks, the lungs were collected and analysed. Picrosirus red staining of lung interstitial collagen content showed that the atherogenic diet increased fibrosis 2.9-fold (P < 0.05), bronchiole adventitial collagen was increased 2.3-fold (P < 0.05) and bronchiole epithelium was increased 34-fold (P < 0.05), and simvastatin treatment severely reduced this effect (P < 0.05). Western blot analysis showed that the atherogenic diet significantly reduced lung Hsp70 protein by 22% (P < 0.05) and Hsp90 protein by 18% (P < 0.05) and simvastatin treatment did not affect this result. However, aortic hyper-responsiveness to vasoconstrictors (angiotensin II and phenylephrine) were markedly reduced by simvastatin treatment. We report that an atherogenic diet stimulates pulmonary fibrosis and reduces lung Hsp70/Hsp90 protein concentration. Simvastatin impairs this by mechanisms unrelated to Hsp70/Hsp90, but possibly a reduction in angiotensin II receptor or alpha adrenergic receptor pathways. These results could have implications in idiopathic pulmonary fibrosis.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%