Merck
  • Loss of miR-200 family in 5-fluorouracil resistant colon cancer drives lymphendothelial invasiveness in vitro.
Loss of miR-200 family in 5-fluorouracil resistant colon cancer drives lymphendothelial invasiveness in vitro.
Human molecular genetics (2015-04-03)
Daniel Senfter, Silvio Holzner, Maria Kalipciyan, Anna Staribacher, Angelika Walzl, Nicole Huttary, Sigurd Krieger, Stefan Brenner, Walter Jäger, Georg Krupitza, Helmut Dolznig, Robert M Mader
ABSTRACT
Invasive colorectal cancer is associated with poor prognosis requiring treatment with systemic chemotherapies usually including 5-fluorouracil. A consequence of prolonged treatment is the acquisition of resistance eventually resulting in the recurrence of highly metastatic cancer cells. To address the relationship between drug resistance and increased lymphatic metastatic potential, we used a 3D co-culture model of colon tumour cell spheroids of parent CCL227 cells and subclones with gradually increasing resistance against 5-fluorouracil. From each investigated cell line, homogeneous tumour spheroids were generated in the presence of methylcellulose yielding emboli of ∼700 µm diameter. When invasive, tumour spheroids disrupt the continuous lymphendothelial cell (LEC) layer and generate a 'circular chemorepellent-induced defect' (CCID), reminiscent of the entry gates through which tumour emboli intravasate lymphatic vasculature. Here we provide evidence that increasingly chemoresistant colon cancer spheroids were strongly associated with enhanced intravasative properties. In naïve CCL227 spheroids, miR-200 family members were released into exosomes thereby repressing the epithelial to mesenchymal transition-regulating transcription factors ZEB1 and SLUG in LEC. As a consequence of attenuated plasticity and migration of LEC, CCID formation was impaired. Loss of exosomal transferred miR-200c in resistant colon cells rendered LEC more susceptible to pro-migratory signals that were generated and directly transmitted by colon cancer spheroids. This observation indicates a common molecular axis in colon cancer and LEC where miR-200 family members act as regulators of ZEB proteins. The data support the notion that horizontal miR-200 signalling prevents the permeation of cells into adjacent epithelia and contributes to organ integrity.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, amber vial color, pack of 100
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, amber vial, pack of 100
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Baicalein, 98%
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 100
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 100