Merck
  • Surface functionalisation of carbon for low cost fabrication of highly stable electrochemical DNA sensors.
Surface functionalisation of carbon for low cost fabrication of highly stable electrochemical DNA sensors.
Biosensors & bioelectronics (2015-04-17)
Ahmed M Debela, Mayreli Ortiz, Valerio Beni, Ciara K O'Sullivan
ABSTRACT
An alternative strategy for surface tethering of DNA probes, where highly reactive glassy carbon (GC) substrates are prepared via electrochemical hydrogenation and electrochemical/chemical chlorination is reported. Thiolated DNA probes and alkanethiols were stably immobilised on the halogenated carbon, with electrochemical chlorination being milder, thus producing less damage to the surface. Electrochemical DNA sensors prepared using this surface chemistry on carbon with electrochemical chlorination providing an improved performance, producing a highly ordered surface and the use of lateral spacers to improve steric accessibility to immobilised probes was not required.
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ruthenium(III) chloride, Ru content 45-55%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Luperox® A98, Benzoyl peroxide, reagent grade, ≥98%
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Luperox® AFR40, Benzoyl peroxide solution, 40 wt. % in dibutyl phthalate
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
6-(Ferrocenyl)hexanethiol