Merck
  • Abortion after deliberate Arthrotec® addition to food. Mass spectrometric detection of diclofenac, misoprostol acid, and their urinary metabolites.
Abortion after deliberate Arthrotec® addition to food. Mass spectrometric detection of diclofenac, misoprostol acid, and their urinary metabolites.
International journal of legal medicine (2014-12-20)
Bernhard Watzer, Klaas J Lusthof, Horst Schweer
ABSTRACT
Arthrotec(®) (AT) is a combination of diclofenac, a nonsteroidal anti-inflammatory drug (NSAID), and misoprostol (MP), a synthetic analogue of prostaglandin E1 (PGE1). MP is a lipophilic methyl ester prodrug. It is readily metabolized to the biologically active misoprostol acid (MPA). During the last few years, medical studies exhibited MP to be an excellent abortive. In this paper, we describe a rare criminal case of MP abortion, initiated by the expectant father. After the abortion, samples of vomit and urine were collected. Systemic exposure to MP is difficult to prove, because both MP and the active metabolite MPA are hardly excreted in urine. Therefore, in addition to routine toxicological analysis, we used slightly modified, well-established liquid and gas chromatographic/tandem mass spectrometric (LC/MS/MS and GC/MS/MS) methods, for the direct and the indirect detection of MPA and its metabolites. In this case, we were able to demonstrate the presence of the major MP metabolites 2,3-dinor-MPA and 2,3,4,5-tetranor-MPA in the urine of the victim. We also detected paracetamol, 3-methoxyparacetamol and diclofenac-glucuronide in the urine. In the vomit of the victim, we detected diclofenac and MPA. These results, combined with the criminal investigations, showed that the accused had mixed MP into the food of his pregnant girlfriend. Finally, these investigations contributed to a confession of the accused.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Diisopropyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diisopropyl ether, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Hydroxylamine hydrochloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube