Skip to Content
Merck
All Photos(1)

Key Documents

612162

Sigma-Aldrich

Phosphoric acid solution

NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

grade

NMR reference standard

Quality Level

Assay

83.5-86.5% (gravimetric)

form

solution

concentration

85% in D2O (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube

density

1.481 g/cm3

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoric acid solution is widely used NMR reference standard.

Application

Phosphoric acid may be employed in the following studies:
  • As standard for the determination of phosphorus-31 nuclear magnetic resonance (31P NMR) chemical shifts of the common phospholipids.[1]
  • As external standard for the analysis of lipid composition of human white matter by 31P NMR spectroscopy.[2]
  • As external standard during the acid-labile phosphorylation of histone H4 by 31P NMR spectroscopy.[3]
  • As reference standard to investigate the surface species present on silica/alumina-supported sodium phosphates by pre- and postreaction MAS31P-NMR and FTIR spectroscopies.[4]

Features and Benefits

31P sensitivity

Packaging

Coaxial 4.2 mm O.D. x 8 in. tube (equivalent to Wilmad WGS-5BL) designed to be used inside a 5 mm x 7 in. NMR tube.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FTIR and 31 P-NMR spectroscopic analyses of surface species in phosphate-catalyzed lactic acid conversion.
Gunter GC, et al.
J. Catal., 164(1), 207-219 (1996)
J M Fujitaki et al.
Biochemistry, 20(12), 3658-3664 (1981-06-09)
Phosphorus-31 nuclear magnetic resonance (31P NMR) is used to investigate acid-labile phosphorylation of histone H4. 31P NMR detects phosphorylated histidine residues in in vitro enzymatically phosphorylated H4. The source of kinase is nuclei from either regenerating rat liver or Walker-256
D S Marinier et al.
Journal of lipid research, 29(9), 1237-1243 (1988-09-01)
In order to study the lipid composition of human white matter, we have developed a 31P NMR spectroscopy method, which allows the determination and quantitation of the main phospholipids found in biological membranes. The technique is based upon the use
W M Fennis et al.
Journal of dental research, 93(1), 36-41 (2013-10-25)
The objective of this randomized control trial was to compare the five-year clinical performance of direct and indirect resin composite restorations replacing cusps. In 157 patients, 176 restorations were made to restore maxillary premolars with Class II cavities and one
Li Yan Chan et al.
The Journal of organic chemistry, 78(17), 8826-8832 (2013-08-07)
A simple and efficient method is developed for Pd-catalyzed ortho-acetoxylation using organophosphates, namely, benzylic phosphonic and aryl phosphoric monoacids, as the directing group.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service