Merck
  • Alpine Skiing With total knee ArthroPlasty (ASWAP): metabolism, inflammation, and skeletal muscle fiber characteristics.
Alpine Skiing With total knee ArthroPlasty (ASWAP): metabolism, inflammation, and skeletal muscle fiber characteristics.
Scandinavian journal of medicine & science in sports (2015-06-18)
M Kristensen, B Pötzelsberger, P Scheiber, A Bergdahl, C N Hansen, J L Andersen, M Narici, S Salvioli, M Conte, E Müller, F Dela
ABSTRACT
We investigated the effect of alpine skiing for 12 weeks on skeletal muscle characteristics and biomarkers of glucose homeostasis and cardiovascular risk factors. Twenty-three patients with a total knee arthroplasty (TKA) were studied 2.9 ± 0.9 years (mean ± SD) after the operation. Fourteen patients participated in the intervention group (IG) and nine in the control group (CG). Blood samples and muscle biopsies were obtained before (PRE) and 7.3 ± 0.8 days after (POST) the intervention, and blood samples again after a retention (RET) phase of 8 weeks. With skiing, glucose homeostasis improved in IG (decrease in fasting insulin, increase in muscle glycogen) but not in CG. Fiber type distribution and size, as well as capillary density and number of capillaries around the fibers (CAF), were not different between the operated and the non-operated leg in either group. The relative number of type I fibers increased with skiing in IG with no change in CG. Inflammatory biomarkers, plasma lipids, and mitochondrial proteins and activity did not change. Alpine skiing is metabolically beneficial and can be used as a training modality by elderly people with TKA.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Lectin from Ulex europaeus (gorse, furze), lyophilized powder
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Anti-Lectin, Ulex europaeus-I antibody produced in rabbit, whole antiserum, delipidized
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%