Merck
  • Kinetics and equilibria of the chromatographic separation of maltose and trehalose.
Kinetics and equilibria of the chromatographic separation of maltose and trehalose.
Journal of separation science (2015-04-16)
Dengjun He, Jiachun Zhou, Quanming Xia, Lihua Jiang, Yongjun Qiu, Liming Zhao
ABSTRACT
Trehalose, a nonreducing disaccharide, has been extensively applied to food, cosmetics, and pharmaceutical goods. The resultant solution of trehalose prepared by enzymatic methods includes high amounts of maltose. However, it is quite difficult to separate maltose and trehalose on an industrial scale because of their similar properties. In this paper, a high-performance resin was selected as a stationary phase to separate trehalose and maltose, and the resolution of these sugars was 0.59. The potential of a cation exchange resin was investigated as the stationary phase in separating trehalose and maltose using deionized water as the mobile phase. Based on the equilibrium dispersive model, the axial dispersion coefficients and overall mass transfer coefficients of maltose and trehalose were determined by moment analysis at two different temperatures, 50 and 70°C. Other parameters, including the column void and the adsorption isotherms, were also determined and applied to simulate the elution curves of trehalose and maltose. The simulated results matched the experimental data, validating the parameters. The optimized parameters are critical to the chromatographic separation of trehalose and maltose on an industrial scale.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
D-(+)-Maltose monohydrate, SAJ special grade, ≥99.0%
Sigma-Aldrich
D-(+)-Maltose monohydrate, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Maltose monohydrate, from potato, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Maltose monohydrate, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(+)-Maltose monohydrate, ≥98% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Maltose monohydrate, Type II, ≥95%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications