Merck
  • Development of an injection molded ethylene-vinyl acetate copolymer (EVA) intravaginal insert for the delivery of progesterone to cattle.
Development of an injection molded ethylene-vinyl acetate copolymer (EVA) intravaginal insert for the delivery of progesterone to cattle.
Animal reproduction science (2015-05-30)
A J Cappadoro, J A Luna
ABSTRACT
The purpose of this study was to develop a new injection-molded intravaginal insert manufactured from ethylene-vinyl acetate containing progesterone for a 7-day insertion period in cattle. The manufacturing process resulted in a reduction in the residual drug compared to the silicone insert available while still maintaining biological performance.
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetaldehyde diethyl acetal, 99%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Acetal, ≥98%, FG
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Sigma-Aldrich
Acetal, natural, ≥97%, FG
Sigma-Aldrich
Vinyl acetate, CP