Skip to Content
Merck
All Photos(1)

Key Documents

W200220

Sigma-Aldrich

Acetal

natural, ≥97%, FG

Synonym(s):

Acetaldehyde diethyl acetal, 1,1-Diethoxyethane, Acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OC2H5)2
CAS Number:
Molecular Weight:
118.17
FEMA Number:
2002
Beilstein:
1098310
EC Number:
Council of Europe no.:
35
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.001
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

vapor density

4.1 (vs air)

vapor pressure

20 mmHg ( 20 °C)
8 mmHg ( 8 °C)

Assay

≥97%

autoignition temp.

446 °F

expl. lim.

10.4 %

refractive index

n20/D 1.379-1.383 (lit.)

bp

102 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; earthy; ethereal; vegetable; sweet

SMILES string

CCOC(C)OCC

InChI

1S/C6H14O2/c1-4-7-6(3)8-5-2/h6H,4-5H2,1-3H3

InChI key

DHKHKXVYLBGOIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<68.0 °F - closed cup

Flash Point(C)

< 20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geelsu Hwang et al.
Biofouling, 30(9), 1079-1091 (2014-10-31)
Well-established biofilms formed by Streptococcus mutans via exopolysaccharide matrix synthesis are firmly attached to tooth surfaces. Enhanced understanding of the physical properties of mature biofilms may lead to improved approaches to detaching or disassembling these highly organized and adhesive structures.
A J Cappadoro et al.
Animal reproduction science, 158, 104-108 (2015-05-30)
The purpose of this study was to develop a new injection-molded intravaginal insert manufactured from ethylene-vinyl acetate containing progesterone for a 7-day insertion period in cattle. The manufacturing process resulted in a reduction in the residual drug compared to the
Federica Sangiuolo et al.
The European respiratory journal, 45(2), 483-490 (2014-12-17)
In idiopathic pulmonary fibrosis (IPF), lung accumulation of excessive extracellular iron and macrophage haemosiderin may suggest disordered iron homeostasis leading to recurring microscopic injury and fibrosing damage. The current study population comprised 89 consistent IPF patients and 107 controls. 54
Mikiko Tsukimoto et al.
Biopharmaceutics & drug disposition, 36(1), 15-33 (2014-09-30)
Aliskiren is a substrate for P-glycoprotein (P-gp) and is metabolized via cytochrome P450 3A4 (CYP3A4). The aim of the present study was to assess whether P-gp influenced the pharmacokinetics of aliskiren and also if drug-drug interactions (DDIs) mediated through P-gp
Sandra N Staeheli et al.
Analytical and bioanalytical chemistry, 407(29), 8681-8712 (2015-09-24)
Postmortem redistribution (PMR) is one of numerous problems in postmortem toxicology making correct interpretation of measured drug concentrations difficult or even impossible. Time-dependent PMR in peripheral blood and especially in tissue samples is still under-explored. For further investigation, an easy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service