Merck
  • Par-3 modulates intestinal epithelial barrier function through regulating intracellular trafficking of occludin and myosin light chain phosphorylation.
Par-3 modulates intestinal epithelial barrier function through regulating intracellular trafficking of occludin and myosin light chain phosphorylation.
Journal of gastroenterology (2015-03-31)
Min Yu, Songwei Yang, Yuan Qiu, Guoqing Chen, Wensheng Wang, Chao Xu, Wenqiang Cai, Lihua Sun, Weidong Xiao, Hua Yang
ABSTRACT
Tight junctions play a critical role in the maintenance of intestinal barrier function. Partitioning-defective protein 3 (Par-3) can regulate intestinal barrier function through the modulation of tight junction assembly and cell polarity. However, the mechanisms are still not fully understood. Adult C57BL/6 mice were treated with dextran sulfate sodium for 7 days, and segments of colon were harvested for immunofluorescent staining of Par-3. Caco-2 intestinal epithelial cells were treated with tumor necrosis factor α (TNF-α) for 24 h, and Par-3 expression was detected by Western blot analysis and immunofluorescence. Additionally, Caco-2 cells were treated with Par-3 small interfering RNA, and altered expression and subcellular localization of tight junction proteins were studied by Western blot analysis and immunofluorescence. Furthermore, the interaction between Par-3 and myosin light chain (MLC) was detected by immunoprecipitation. Par-3 was downregulated in murine dextran sulfate sodium induced acute inflammation and TNF-α-treated Caco-2 cells. Depletion of Par-3 expression by small interfering RNA delayed intestinal epithelial barrier development in Caco-2 cells. This regulation was due to the redistribution of the tight junction protein occludin rather than the altered total levels of tight junction proteins. Par-3 silencing blocked the trafficking of occludin from or through the Golgi complex to the cell surface, and dramatically induced occludin accumulated at the Golgi complex. Importantly, Par-3 can interact with MLC, and loss of Par-3 upregulated MLC kinase expression and MLC phosphorylation, which contributed to intestinal epithelial barrier dysfunction. These results indicate that Par-3 plays an important role in the modulation of intestinal barrier function by regulating delivery of occludin as well as suppression of MLC phosphorylation.
Sigma-Aldrich
MISSION® esiRNA, targeting human OCLN
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Trizma® hydrochloride, BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline