Merck
  • Synthesis and characterization of bismuth(III) and antimony(V) porphyrins: high antileishmanial activity against antimony-resistant parasite.
Synthesis and characterization of bismuth(III) and antimony(V) porphyrins: high antileishmanial activity against antimony-resistant parasite.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2015-05-02)
Marcela Luísa Gomes, Gilson DeFreitas-Silva, Priscila Gomes dos Reis, Maria Norma Melo, Frédéric Frézard, Cynthia Demicheli, Ynara Marina Idemori
ABSTRACT
Two bismuth(III) porphyrins-5,10,15,20-tetrakis(phenyl)porphyrinatobismuth(III) nitrate, [Bi(III)(TPP)]NO3, and the unprecedent 5,10,15,20-tetrakis(4-carbomethoxyphenyl)porphyrinatobismuth(III) nitrate, [Bi(III)(T4CMPP)]NO3, and two unprecedented antimony(V) porphyrins dichlorido(5,10,15,20-tetrakis(phenyl)porphyrinato)antimony(V) bromide, [Sb(V)(TPP)Cl2]Br, and dibromido(5,10,15,20-tetrakis(4-carbomethoxyphenyl)porphyrinato)antimony(V) bromide, [Sb(V)(T4CMPP)Br2]Br,-were synthesized by reacting the corresponding porphyrin ligand with Bi(NO3)3·5H2O or SbCl3. All compounds were characterized by UV-vis, (1)H NMR spectroscopy, and mass spectrometry. The new compounds were also characterized by elemental analysis. Because antimony and bismuth compounds have been widely applied in medicine, the activity of these complexes was tested against Sb-sensitive and -resistant Leishmania amazonensis parasites. [Sb(V)(T4CMPP)Br2]Br was more active against the promastigote form of Sb-resistant mutant strain as compared to the sensitive parental strain, with IC50 in the micromolar range. These data contrasted with those obtained using the Sb(III) drug potassium antimony tartrate, which displayed IC50 of 110 μmol L(-1) against the Sb-sensitive parasite and was almost inactive against the Sb-resistant strain. The H2T4CMPP ligand also showed antileishmanial activity against Sb-resistant and -sensitive strains, but with IC50 at least tenfold greater than that of the complex. The Sb(V)-porphyrin complex was also active against intracellular amastigotes and showed a higher selectivity index than the conventional Sb(V) drug glucantime, in both Sb-sensitive and -resistant strains. The greater antileishmanial activity of this complex could be attributed to an increased cellular uptake of Sb. Thus, [Sb(V)(T4CMPP)Br2]Br constitutes a new antileishmanial drug candidate.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Antimony(III) chloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, reagent grade, 98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Antimony(III) chloride, ≥99.95% trace metals basis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%