Merck
  • Evaluating the thermal vinylcyclopropane rearrangement (VCPR) as a practical method for the synthesis of difluorinated cyclopentenes: experimental and computational studies of rearrangement stereospecificity.
Evaluating the thermal vinylcyclopropane rearrangement (VCPR) as a practical method for the synthesis of difluorinated cyclopentenes: experimental and computational studies of rearrangement stereospecificity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-09-13)
David Orr, Jonathan M Percy, Tell Tuttle, Alan R Kennedy, Zoë A Harrison
ABSTRACT
Vinyl cyclopropane rearrangement (VCPR) has been utilised to synthesise a difluorinated cyclopentene stereospecifically and under mild thermal conditions. Difluorocyclopropanation chemistry afforded ethyl 3-(1'(2'2'-difluoro-3'-phenyl)cyclopropyl) propenoate as all four stereoisomers (18a, 18b, 22a, 22b) (all racemic). The trans-E isomer (18a), prepared in 70 % yield over three steps, underwent near quantitative VCPR to difluorocyclopentene 23 (99 %). Rearrangements were monitored by (19) F NMR (100-180 °C). While cis/trans cyclopropane stereoisomerisation was facile, favouring trans-isomers by a modest margin, no E/Z alkene isomerisation was observed even at higher temperatures. Neither cis nor trans Z-alkenoates underwent VCPR, even up to much higher temperatures (180 °C). The cis-cyclopropanes underwent [3,3]-rearrangement to afford benzocycloheptadiene species. The reaction stereospecificity was explored by using electronic structure calculations, and UB3LYP/6-31G* methodology allowed the energy barriers for cyclopropane stereoisomerisation, diastereoisomeric VCPR and [3,3]-rearrangement to be ranked in agreement with experiment.
Sigma-Aldrich
Potassium iodide, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium iodide, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%