Skip to Content
Merck
All Photos(1)

Key Documents

60422

Sigma-Aldrich

Potassium iodide

tested according to Ph. Eur.

Synonym(s):

Kalii iodidum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
solid
solubility:
H2O: soluble 1 M at 20 °C, colorless (completely)

grade

for analytical purposes

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

vapor pressure

1 mmHg ( 745 °C)

form

solid

mp

681 °C (lit.)

solubility

H2O: soluble 1 M at 20 °C, colorless (completely)

storage temp.

2-8°C

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Decorporation dilemma: Interplay of prussian blue and potassium iodide in radioactive contamination.: This review discusses the combined use of prussian blue and potassium iodide for radioactive decontamination, highlighting their synergistic effects and potential for enhanced efficacy in removing radioactive substances from the body (Mahar et al., 2024).

  • Tooth Discoloration from 2 Silver Fluorides Used in Adults with Special Needs: A Randomized Trial.: This research evaluates the effects of potassium iodide in conjunction with silver fluoride on tooth discoloration, providing insights into its potential benefits and drawbacks in dental treatments for special needs patients (See et al., 2024).

  • A cost-effective and facile technique for realizing fabric-based microfluidic channels using beeswax and PVC stencils.: The paper describes a novel method for fabricating microfluidic channels using potassium iodide, showcasing its utility in creating precise and cost-effective microfluidic devices (P et al., 2024).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Santulli et al.
Human reproduction (Oxford, England), 30(1), 49-60 (2014-11-08)
Are protein oxidative stress markers [thiols, advanced oxidation protein products (AOPP), protein carbonyls and nitrates/nitrites] in perioperative peritoneal fluid higher in women with histologically proven endometriosis when compared with endometriosis-free controls? Protein oxidative stress markers are significantly increased in peritoneal
Myrick C Shinall et al.
Annals of surgical oncology, 20(9), 2964-2967 (2013-07-13)
Potassium iodide (KI) has traditionally been used to reduce gland vascularity and diminish blood loss in patients undergoing thyroidectomy for Graves disease (GD). Current American Thyroid Association (ATA) guidelines (Recommendation 22) call for its routine administration in GD but avoidance
H M D Agbogun et al.
Journal of contaminant hydrology, 145, 44-53 (2013-01-10)
X-ray micro-computed tomography (micro-CT) techniques for measuring the three-dimensional (3-D) distributions of diffusion-accessible porosity (φ(d)) and temporal tracer-concentrations (C(t)) within a dolostone sample subjected to solute diffusion are developed and tested in this work. The φ(d) and C(t) measurements are
Zhi-Qi Lao et al.
Organic & biomolecular chemistry, 10(39), 7869-7871 (2012-09-06)
We have developed a new KI-catalyzed method for the imidation of an sp(3) C-H bond adjacent to an amide nitrogen atom by using TBHP (tert-butyl hydroperoxide, 70% aqueous solution) as the oxidant. This novel procedure tolerated air and moisture and
Juan Pablo Nicola et al.
The Journal of physiology, 590(23), 6013-6026 (2012-09-26)
Dietary I(-) absorption in the gastrointestinal tract is the first step in I(-) metabolism. Given that I(-) is an essential constituent of the thyroid hormones, its concentrating mechanism is of significant physiological importance. We recently described the expression of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service