Merck
  • Crucial roles of interleukin-7 in the development of T follicular helper cells and in the induction of humoral immunity.
Crucial roles of interleukin-7 in the development of T follicular helper cells and in the induction of humoral immunity.
Journal of virology (2014-06-06)
Yong Bok Seo, Se Jin Im, Hong Namkoong, Sae Won Kim, Young Woo Choi, Moon Cheol Kang, Hye Seong Lim, Hyun Tak Jin, Se Hwan Yang, Mi La Cho, You-Me Kim, Seung-Woo Lee, Young Ki Choi, Charles D Surh, Young Chul Sung
ABSTRACT
T follicular helper (Tfh) cells are specialized providers of cognate B cell help, which is important in promoting the induction of high-affinity antibody production in germinal centers (GCs). Interleukin-6 (IL-6) and IL-21 have been known to play important roles in Tfh cell differentiation. Here, we demonstrate that IL-7 plays a pivotal role in Tfh generation and GC formation in vivo, as treatment with anti-IL-7 neutralizing antibody markedly impaired the development of Tfh cells and IgG responses. Moreover, codelivery of mouse Fc-fused IL-7 (IL-7-mFc) with a vaccine enhanced the generation of GC B cells as well as Tfh cells but not other lineages of T helper cells, including Th1, Th2, and Th17 cells. Interestingly, a 6-fold-lower dose of an influenza virus vaccine codelivered with Fc-fused IL-7 induced higher antigen-specific and cross-reactive IgG titers than the vaccine alone in both mice and monkeys and led to markedly enhanced protection against heterologous influenza virus challenge in mice. Enhanced generation of Tfh cells by IL-7-mFc treatment was not significantly affected by the neutralization of IL-6 and IL-21, indicating an independent role of IL-7 on Tfh differentiation. Thus, IL-7 holds promise as a critical cytokine for selectively inducing Tfh cell generation and enhancing protective IgG responses. Here, we demonstrate for the first time that codelivery of Fc-fused IL-7 significantly increased influenza virus vaccine-induced antibody responses, accompanied by robust expansion of Tfh cells and GC B cells as well as enhanced GC formation. Furthermore, IL-7-mFc induced earlier and cross-reactive IgG responses, leading to striking protection against heterologous influenza virus challenge. These results suggest that Fc-fused IL-7 could be used for inducing strong and cross-protective humoral immunity against highly mutable viruses, such as HIV and hepatitis C virus, as well as influenza viruses.
Sigma-Aldrich
IL-7 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Human IL-7 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse IL-7 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%