Skip to Content
Merck
All Photos(2)

Documents

A6435

Sigma-Aldrich

Aluminum potassium sulfate dodecahydrate

BioReagent, suitable for insect cell culture

Synonym(s):

Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
474.39
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

product line

BioReagent

Quality Level

Assay

≥98%

form

powder

technique(s)

cell culture | insect: suitable

mp

92 °C (lit.)

density

1.757 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum potassium sulfate dodecahydrate has been used as an adjuvant of ovalbumin to induce anaphylaxis in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Epinephrine inhibits vascular hyperpermeability during platelet-activating factor-or ovalbumin-induced anaphylaxis
Ma, X, et al.
Royal Society of Chemistry Advances, 7(83), 52762-52771 (2017)
Aaron S Schwartz-Duval et al.
Nature communications, 11(1), 4530-4530 (2020-09-12)
Various cancer cells have been demonstrated to have the capacity to form plasmonic gold nanoparticles when chloroauric acid is introduced to their cellular microenvironment. But their biomedical applications are limited, particularly considering the millimolar concentrations and longer incubation period of
Saya Moriyama et al.
The Journal of experimental medicine, 211(7), 1297-1305 (2014-06-11)
Follicular helper T (Tfh) cells access the B cell follicle to promote antibody responses and are particularly important for germinal center (GC) reactions. However, the molecular mechanisms of how Tfh cells are physically associated with GCs are incompletely understood. We
Gareth R Williams et al.
The Journal of experimental medicine, 211(6), 1019-1025 (2014-05-07)
There is currently no paradigm in immunology that enables an accurate prediction of how the immune system will respond to any given agent. Here we show that the immunological responses induced by members of a broad class of inorganic crystalline
Yan-Feng Li et al.
Nature communications, 5, 4273-4273 (2014-07-01)
Germline or B-cell-specific loss of Ptpn6 gene encoding the Shp1 protein tyrosine phosphatase leads to skewed B lymphopoiesis and systemic autoimmunity. Here, to study its role in B-cell terminal differentiation, we generated Ptpn6(f/f)Aicda(Cre/+) mice with Shp1 ablated only in activated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service