Skip to Content
Merck
All Photos(3)

Documents

A7210

Sigma-Aldrich

Aluminum potassium sulfate dodecahydrate

BioXtra, ≥98.0%

Synonym(s):

Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
474.39
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥98.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

mp

92 °C (lit.)

solubility

H2O: 0.5 M, clear, colorless

density

1.757 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.001%
Mg: ≤0.005%
NH4+: ≤0.005%
Na: ≤0.02%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saya Moriyama et al.
The Journal of experimental medicine, 211(7), 1297-1305 (2014-06-11)
Follicular helper T (Tfh) cells access the B cell follicle to promote antibody responses and are particularly important for germinal center (GC) reactions. However, the molecular mechanisms of how Tfh cells are physically associated with GCs are incompletely understood. We
Gareth R Williams et al.
The Journal of experimental medicine, 211(6), 1019-1025 (2014-05-07)
There is currently no paradigm in immunology that enables an accurate prediction of how the immune system will respond to any given agent. Here we show that the immunological responses induced by members of a broad class of inorganic crystalline
Yan-Feng Li et al.
Nature communications, 5, 4273-4273 (2014-07-01)
Germline or B-cell-specific loss of Ptpn6 gene encoding the Shp1 protein tyrosine phosphatase leads to skewed B lymphopoiesis and systemic autoimmunity. Here, to study its role in B-cell terminal differentiation, we generated Ptpn6(f/f)Aicda(Cre/+) mice with Shp1 ablated only in activated
Ina Callebaut et al.
American journal of rhinology & allergy, 28(4), 290-296 (2014-09-10)
The naso-ocular interaction in allergic rhinoconjunctivitis is well recognized from epidemiological, clinical, and experimental observations. The precise mechanisms remain incompletely understood. A new mouse model of allergic rhinoconjunctivitis was used to investigate the contribution of mast cells and trigeminal ganglia
Upasana Ray et al.
Science translational medicine, 7(306), 306ra151-306ra151 (2015-09-25)
JC polyomavirus (JCV) persistently infects the urinary tract of most adults. Under conditions of immune impairment, JCV causes an opportunistic brain disease, progressive multifocal leukoencephalopathy (PML). JCV strains found in the cerebrospinal fluid of PML patients contain distinctive mutations in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service