Merck
  • Modulation of tissue inflammatory response by histamine receptors in scorpion envenomation pathogenesis: involvement of H4 receptor.
Modulation of tissue inflammatory response by histamine receptors in scorpion envenomation pathogenesis: involvement of H4 receptor.
Inflammation (2014-05-27)
Amal Lamraoui, Sonia Adi-Bessalem, Fatima Laraba-Djebari
ABSTRACT
The inflammatory response caused by scorpion venoms is a key event in the pathogenesis of scorpion envenomation. This response was assessed in the cardiac, pulmonary, and gastric tissues of envenomed mice. The results reveal an increase of permeability in cardiac, pulmonary, and gastric vessels accompanied by an edema-forming, inflammatory cell infiltration, and imbalanced redox status. These effects are correlated with severe tissue alterations and concomitant increase of metabolic enzymes in sera. Pretreatment of mice with antagonists of H1, H2, or H4 receptors markedly alleviated these alterations in the heart and lungs. Nevertheless, the blockade of the H3 receptor slightly reduced these disorders. Histamine H2 and H4 receptors were the most pharmacological targets involved in the gastric oxidative inflammation. These findings could help to better understand the role of histamine in scorpion venom-induced inflammatory response and propose new therapy using as targets the H4 receptor in addition to histamine H1 and H2 receptors to attenuate the induced inflammatory disorders encountered in scorpion envenoming.
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, ≥99.7%
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Cimetidine
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Cimetidine, European Pharmacopoeia (EP) Reference Standard
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard