Skip to Content
Merck
All Photos(4)

Key Documents

T9159

Sigma-Aldrich

Trichloroacetic acid

BioXtra, ≥99.0%

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein:
970119
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NB.22

grade

BioPerformance Certified

Quality Level

vapor density

<1 (vs air)

vapor pressure

1 mmHg ( 51 °C)

product line

BioXtra

Assay

≥99.0%

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.03%

refractive index

n20/D 1.62 (lit.)

bp

196 °C (lit.)

mp

54-58 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

1.62 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloroacetic acid (TCA) is an important metabolic product of trichloroethylene (TCE) metabolism.

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.
Trichloroacetic acid has been used:
  • for protein precipitation
  • as a component in the extract solution used for the measurement of malondialdehyde and glutathione
  • to precipitate the transfected and treated cells in [3H]-thymidine incorporation assay

Biochem/physiol Actions

TCA might act as a carcinogen, targeting liver. It might be used as an acid decalcifying agent. TCA helps in nuclear staining.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid analysis of total plasma homocysteine by tandem mass spectrometry
Tuschl K, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 351(1-2), 139-141 (2005)
Circulating fibrocytes stabilize blood vessels during angiogenesis in a paracrine manner
Li J, et al.
The American Journal of Pathology, 184(2), 556-571 (2014)
Histophatologic Techniques (2006)
Analysis of malondialdehyde, chlorophyll proline, soluble sugar, and glutathione content in Arabidopsis seedling
Zhang Z and Huang R
Bio-protocol, 3, e817-e817 (2013)
Bo Chen et al.
Translational stroke research, 10(1), 91-103 (2018-03-24)
The transient receptor potential melastatin 4 (TRPM4) channel has been suggested to play a key role in the treatment of ischemic stroke. However, in vivo evaluation of TRPM4 channel, in particular by direct channel suppression, is lacking. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service