Merck
  • Qualitative screening of 116 veterinary drugs in feed by liquid chromatography-high resolution mass spectrometry: potential application to quantitative analysis.
Qualitative screening of 116 veterinary drugs in feed by liquid chromatography-high resolution mass spectrometry: potential application to quantitative analysis.
Food chemistry (2014-05-07)
Clara Boix, María Ibáñez, Juan V Sancho, Nuria León, Vicent Yusá, Félix Hernández
ABSTRACT
Veterinarian and human pharmaceuticals may be intentionally added to animal feed to enhance animal production. Monitoring these substances is necessary for protecting the consumers. In this work, a screening method covering 116 human and veterinary drugs has been developed and validated in five types of animal feed at 0.02 and 0.2 mg kg(-1). After a simple extraction and dilution, the samples were analysed by ultra-high performance liquid chromatography coupled to quadrupole time-of-flight mass spectrometry (UHPLC-QTOF MS). Nearly all compounds tested were detected at 0.02 mg kg(-1), based on the presence of the accurate-mass (de)protonated molecule. However, the identification using a second accurate-mass ion was more problematic at this level. Finally, the procedure was applied to 22 feed samples, where trimethoprim, robenidine, or α- and β-nandrolone were detected and identified. The potential applicability of the method to quantitative analysis of the compounds detected in the samples was also evaluated.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
DNC-d8, VETRANAL®, analytical standard
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, powder
Leucine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard