Merck
  • Study of the persistence of the anti-inflammatory effect observed after application of preparations containing organic ultraviolet filters.
Study of the persistence of the anti-inflammatory effect observed after application of preparations containing organic ultraviolet filters.
International journal of pharmaceutics (2014-10-01)
C Couteau, C Chauvet, E Paparis, L J M Coiffard
ABSTRACT
Given that previous studies have highlighted the anti-inflammatory nature, which is influenced by UV radiation, of a certain number of ultraviolet filters currently used in Europe, it seemed interesting to evaluate the persistence of this type of effect. The persistence of the anti-inflammatory effect of fourteen preparations, each containing one of the ultraviolet filters authorized by Regulation (CE) No. 1223/2009, and of 10 commercially-available sun products was studied using the phorbol-myristate-acetate test on mice, up to 6 and a half hours after application. We can observe that a benzophenone, oxybenzone, a PABA derivative, octyldimethylPABA and a derivative of cinnamic acid, OMC as well as 3 commercially-available products, display a very marked anti-inflammatory effect at the end of our experimentation phase. The type of effect observed could encourage users to prolong the time that they are exposed to the sun, due to there being no warning signs, namely sunburn.
Product Description
Supelco
Oxybenzone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 97%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Trolamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Supelco
Oxybenzone, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
USP
Oxybenzone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Sigma-Aldrich
Titanium(IV) oxide, JIS special grade, ≥99.0%
Supelco
Octocrylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Octocrylene, analytical standard
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)