Skip to Content
Merck
All Photos(1)

Key Documents

415820

Sigma-Aldrich

2-Ethylhexyl 2-cyano-3,3-diphenylacrylate

97%

Synonym(s):

2-Cyano-3,3-diphenylacrylic acid 2-ethylhexyl ester, 2-Ethylhexyl α-cyano-β-phenylcinnamate, 2-Ethylhexyl 2-cyano-3,3-diphenyl-2-propenoate, Octocrylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C=C(CN)CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
361.48
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

viscous liquid

refractive index

n20/D 1.567 (lit.)

bp

218 °C/1.5 mmHg (lit.)

mp

−10 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)\C(C#N)=C(/c1ccccc1)c2ccccc2

InChI

1S/C24H27NO2/c1-3-5-12-19(4-2)18-27-24(26)22(17-25)23(20-13-8-6-9-14-20)21-15-10-7-11-16-21/h6-11,13-16,19H,3-5,12,18H2,1-2H3

InChI key

FMJSMJQBSVNSBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexyl 2-cyano-3,3-diphenylacrylate is an ester-based UV filter formed by reacting 3,3-diphenylcyanoacrylate and 2-ethylhexanol. It is commercially used as a UV absorber in sunscreens and other cosmetic formulations.

Application

  • Photochemical response of the scleractinian coral Stylophora pistillata to some sunscreen ingredients: This study investigates the impact of octocrylene on the photosynthetic efficiency of coral symbionts, raising concerns about environmental effects (JP Fel et al., 2019).
  • Preparation of O/I1-type emulsions and S/I1-type dispersions encapsulating UV-absorbing agents: Explores formulations for incorporating octocrylene into cosmetic products, enhancing UV protection capabilities (K Aramaki et al., 2015).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

453.2 °F - closed cup

Flash Point(C)

234 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive solid-phase extraction based on oleic acid-coated magnetic nanoparticles followed by gas chromatography-mass spectrometry for UV-filter determination in water samples
Roman IP, et al.
Journal of Chromatography A, 1218(18), 2467-2475 (2011)
Changes in ultraviolet absorbance and hence in protective efficacy against lipid peroxidation of organic sunscreens after UVA irradiation
Damiani E, et al.
Journal of Photochemistry and Photobiology. B, Biology, 82(3), 204-213 (2006)
Sunscreen enhancement of UV-induced reactive oxygen species in the skin
Hanson KM, et al.
Free Radical Biology & Medicine, 41(8), 1205-1212 (2006)
Weihong Li et al.
Water research, 41(15), 3506-3512 (2007-06-29)
Four UV filters, benzophenone-3 (BP-3), 4-methylbenzylidence camphor (4-MBC), ethylhexyl methoxycinnamate (EHMC), and octocrylene (OC), have been examined along the different units of a wastewater reclamation plant (WWRP) located in Tianjin, North China. The analytical procedure included solid-phase extraction and gas
N Negreira et al.
Analytical and bioanalytical chemistry, 400(2), 603-611 (2011-02-22)
This work describes an effective, low solvent consumption and affordable sample preparation approach for the determination of eight UV filters in surface and wastewater samples. It involves sorptive extraction of target analytes in a disposable, technical grade silicone disc (5 mm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service