Merck
  • Human oocyte-derived sperm chemoattractant is a hydrophobic molecule associated with a carrier protein.
Human oocyte-derived sperm chemoattractant is a hydrophobic molecule associated with a carrier protein.
Fertility and sterility (2014-07-27)
Leah Armon, Ido Ben-Ami, Raphael Ron-El, Michael Eisenbach
ABSTRACT
To characterize the nature of the human oocyte-derived chemoattractant. Laboratory in vitro study. Academic research institute. Ten healthy sperm donors. Oocyte-conditioned media from women undergoing IVF treatment because of male factor infertility. Sperm samples were processed by the migration-sedimentation technique. Oocyte-conditioned media were collected 2-3 hours after oocyte stripping. Sperm chemotaxis was assayed in a μ-slide chamber according to the direction of swimming relative to that of the chemical gradient. Oocyte-conditioned media treated with proteases did not lose their chemotactic activity; on the contrary, they became more active, with the activity shifted to lower concentrations. When oocyte-conditioned media were subjected to hexane extraction, chemotactic activity was found in both the hydrophobic and aqueous phases. Known mammalian sperm chemoattractants were ruled out as oocyte-derived chemoattractants. Our results suggest that the oocyte-derived chemoattractant is a hydrophobic nonpeptide molecule that, in an oocyte-conditioned medium, is associated with a carrier protein that enables its presence in a hydrophilic environment.
Product Description
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Progesterone, ≥99%
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Supelco
Undecanal, analytical standard
Alprostadil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)