Merck
  • Effect of co-ligands on chemical and biological properties of (99m)Tc(III) complexes [(99m)Tc(L)(CDO)(CDOH)2BMe] (L=Cl, F, SCN and N3; CDOH2=cyclohexanedione dioxime).
Effect of co-ligands on chemical and biological properties of (99m)Tc(III) complexes [(99m)Tc(L)(CDO)(CDOH)2BMe] (L=Cl, F, SCN and N3; CDOH2=cyclohexanedione dioxime).
Nuclear medicine and biology (2014-08-30)
Yumin Zheng, Shundong Ji, Elena Tomaselli, Carley Ernest, Tom Freiji, Shuang Liu
ABSTRACT
(99m)Tc-Teboroxime ([(99m)TcCl(CDO)(CDOH)2BMe]) is a member of the BATO (boronic acid adducts of technetium dioximes) class of (99m)Tc(III) complexes. This study sought to explore the impact of co-ligands on solution stability, heart uptake and myocardial retention of [(99m)Tc(L)(CDO)(CDOH)2BMe] ((99m)Tc-Teboroxime: L=Cl; (99m)Tc-Teboroxime(F): L=F; (99m)Tc-Teboroxime(SCN): L=SCN; and (99m)Tc-Teboroxime(N3): L=N3). Radiotracers (99m)Tc-Teboroxime(L) (L=F, SCN and N3) were prepared by reacting (99m)Tc-Teboroxime with NaF, NaSCN and NaN3, respectively. Biodistribution and imaging studies were carried out in Sprague-Dawley rats. Image quantification was performed to compare their heart retention and liver clearance kinetics. Complexes (99m)Tc-Teboroxime(L) (L=F, SCN and N3) were prepared in high yield with high radiochemical purity. All new radiotracers were stable for >6h in the kit matrix. In its HPLC chromatogram, (99m)Tc-Teboroxime showed one peak at ~15.5 min, which was shorter than that of (99m)Tc-Teboroxime(F) (~16.4 min). There were two peaks for (99m)Tc-Teboroxime(SCN) at 16.5 and 18.3 min. (99m)Tc-Teboroxime(N3) appeared as a single peak at 18.4 min. Their heart retention and liver clearance curves were best fitted to the bi-exponential decay function. The half-times of fast/slow components were 1.6±0.4/60.7±8.9 min for (99m)Tc-Teboroxime, 0.8±0.2/101.7±20.7 min for (99m)Tc-Teboroxime(F), 1.2±0.3/84.8±16.6 min for (99m)Tc-Teboroxime(SCN), and 2.9±0.9/51.6±5.0 min for (99m)Tc-Teboroxime(N3). The 2-min heart uptake followed the order of (99m)Tc-Teboroxime (3.00±0.37%ID/g)>(99m)Tc-Teboroxime(N3) (2.66±0.01 %ID/g)≈(99m)Tc-Sestamibi (2.55±0.46 %ID/g)>(99m)TcN-MPO (2.38±0.15 %ID/g). (99m)Tc-Teboroxime remains the best in first-pass extraction. The best image acquisition window is 0-5 min for (99m)Tc-Teboroximine and 0-15 min for (99m)Tc-Teboroximine(N3). Co-ligands had significant impact on the heart uptake and myocardial retention of complexes [(99m)Tc(L)(CDO)(CDOH)2BMe] (L=Cl, F, SCN and N3). Future studies should be directed towards minimizing the liver uptake and radioactivity accumulation in the blood vessels while maintaining their high heart uptake.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
1,2-Cyclohexanedione dioxime, 96%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O