Merck
  • Binding to large enzyme pockets: small-molecule inhibitors of trypanothione reductase.
Binding to large enzyme pockets: small-molecule inhibitors of trypanothione reductase.
ChemMedChem (2014-05-03)
Elke Persch, Steve Bryson, Nickolay K Todoroff, Christian Eberle, Jonas Thelemann, Natalie Dirdjaja, Marcel Kaiser, Maria Weber, Hassan Derbani, Reto Brun, Gisbert Schneider, Emil F Pai, R Luise Krauth-Siegel, François Diederich
ABSTRACT
The causative agents of the parasitic disease human African trypanosomiasis belong to the family of trypanosomatids. These parasitic protozoa exhibit a unique thiol redox metabolism that is based on the flavoenzyme trypanothione reductase (TR). TR was identified as a potential drug target and features a large active site that allows a multitude of possible ligand orientations, which renders rational structure-based inhibitor design highly challenging. Herein we describe the synthesis, binding properties, and kinetic analysis of a new series of small-molecule inhibitors of TR. The conjunction of biological activities, mutation studies, and virtual ligand docking simulations led to the prediction of a binding mode that was confirmed by crystal structure analysis. The crystal structures revealed that the ligands bind to the hydrophobic wall of the so-called "mepacrine binding site". The binding conformation and potency of the inhibitors varied for TR from Trypanosoma brucei and T. cruzi.
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, analytical standard
Supelco
Toluene, HPLC grade, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%