Merck
Intramolecular Hydrogen Bonding and Linear Pentapyrrole Conformation.
Monatshefte fur chemie (2014-08-12)
D Timothy Anstine, David A Lightner
ABSTRACT
Three new linear pentapyrrole rubinoid analogs: 2,3,7,8,17,18,22,23-octamethyl-12,13-bis-(2-carboxyethyl)-1,10,15,24,25,27,28,29-octahydro-27H-pentapyrrin-1,24-dione and 2,3,8,12,13,17,22,23-octamethyl-7,18-bis-(2-carboxyethyl)-1,10,15,24,25,26,27,28,29-octahydro-27H-pentapyrrin-1,24-dione, and its 7,18-dihexanoic acid analog were synthesized, respectively, from 2,3,7,8-tetramethyl-(10H)-dipyrrin-2-one, from 2,3,8-trimethyl-7-[2-(methoxycarbonyl)ethyl]-(10H)-dipyrrinone, and 2,3,8-trimethyl-7-[5-(methoxycarbonyl)pentyl]-(10H)-dipyrrinone. 13C NMR and 1H NMR measurements in (CD3)2SO confirmed the pentapyrrole structures, while 1H NMR data indicate intramolecular hydrogen bonding between the CO2H and dipyrrinone groups. Molecular mechanics modeling studies suggest stable U-shape conformations capable of encapsulating small planar aromatic molecules.
Sigma-Aldrich
Ethyl acrylate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
Ethyl cyanoacetate, ≥98%
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized
Sigma-Aldrich
Ethyl 6-bromohexanoate, 99%
Sigma-Aldrich
Trimethyl orthoformate, 99%
Sigma-Aldrich
Trimethyl orthoformate, anhydrous, 99.8%
Sigma-Aldrich
Succinic anhydride, SAJ first grade, ≥99.0%
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Diethyl malonate, ≥98%, FG
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%