Skip to Content
Merck
All Photos(1)

Key Documents

09-0750

Sigma-Aldrich

Ethyl acrylate

SAJ first grade, ≥99.0%

Synonym(s):

Acrylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOC2H5
CAS Number:
Molecular Weight:
100.12
Beilstein:
773866
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

721 °F

expl. lim.

12.1 %

availability

available only in Japan

refractive index

n20/D 1.406 (lit.)

bp

99 °C (lit.)

mp

−71 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3

InChI key

JIGUQPWFLRLWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna E V Hagström et al.
Biotechnology and bioengineering, 102(3), 693-699 (2008-09-30)
An OH-functional polyester has been acrylated via transesterification of ethyl acrylate, catalyzed by Candida antarctica lipase B (CalB) in two different preparations: Novozym 435 and immobilized on Accurel MP1000. The batch process resulted in incomplete acrylation as well as severe
Junliang Wu et al.
Journal of the American Chemical Society, 131(39), 13888-13889 (2009-09-15)
The direct cross-coupling of quinoline-N-oxides with olefin derivatives has been realized using palladium acetate as the catalyst in the absence of external ligand and oxidant to give the corresponding 2-alkenylated quinolines and 1-alkenylated isoquinolines chemo- and regioselectively in 27-95% yield.
M Shahjahan Kabir et al.
The Journal of organic chemistry, 77(1), 300-310 (2011-11-15)
The stereospecific synthesis of aryloxy and amino substituted E- and Z-ethyl-3-acrylates is of interest because of their potential in the polymer industry and in medicinal chemistry. During work on a copper-catalyzed cross-coupling reaction of ethyl (E)- and (Z)-3-iodoacrylates with phenols
M Monier et al.
Journal of hazardous materials, 176(1-3), 348-355 (2009-12-08)
The graft copolymerization of ethyl acrylate (EA) onto natural wool fibers initiated by potassium persulphate and Mohr's salt redox initiator system in limited aqueous medium was carried out in heterogeneous media. Ester groups of the grafted copolymers were partially converted
Aaron Mark Drucker et al.
Dermatitis : contact, atopic, occupational, drug, 22(2), 98-101 (2011-04-21)
Acrylates are present in a wide variety of products and cause occupational and non-occupational allergic contact dermatitis. There is no clear guidance from the literature as to which allergens should be used for patch-test screening for acrylates. To characterize patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service