Skip to Content
Merck
All Photos(1)

Key Documents

537594

Sigma-Aldrich

tert-Butyl acetoacetate

reagent grade, 98%

Synonym(s):

TBAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC(CH3)3
CAS Number:
Molecular Weight:
158.19
Beilstein:
1680303
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

98%

expl. lim.

8 % (lit.)

impurities

≤0.20% water
water

refractive index

n20/D 1.419 (lit.)

bp

71-72 °C/11 mmHg (lit.)

mp

-38 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CC(=O)CC(=O)OC(C)(C)C

InChI

1S/C8H14O3/c1-6(9)5-7(10)11-8(2,3)4/h5H2,1-4H3

InChI key

JKUYRAMKJLMYLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl acetoacetate (t-BAA) is a cheap and easy to store commercial reagent. Reaction demonstrated that the more hindered tert-butyl acetoacetate (t-BAA, la) is ca.15-20-fold more reactive than the more commonly used methyl or ethyl analogs. It is widely employed as an acetoacetylating reagent.

Application

tert-Butyl acetoacetate may be used in the synthesis of:
  • various acetoacetic acid derivatives
  • acetoacetates
  • acetoacetamides
  • 1-(diethylamino)-2-acetoacetoxypropane
  • (S)-tert-butyl 3-hydroxybutyrate
  • benzothiazole β-keto ester derivatives
  • 3,4-disubstituted pyrroles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transesterification.
Otera J.
Chemical Reviews, 93(4), 1449-1470 (1993)
Deborah A Smithen et al.
Organic & biomolecular chemistry, 11(1), 62-68 (2012-10-17)
Prodigiosenes, possessing a 4-methoxypyrrolyldipyrrin skeleton, are known for their anti-cancer activity. Structural modification of the C-ring resulted in a series of prodigiosenes that displayed promising activity against leukemia cell lines during in vitro analysis against the NCI 60 cancer cell
Highly enantioselective Mannich reactions of imines with tert-butyl acetoacetate catalyzed by squaramide organocatalyst.
He HX and Du DM.
Tetrahedron Asymmetry, 25(8), 637-643 (2014)
Enzymatic Synergism in the Synthesis of ?-Keto Esters.
Wisniewska C, et al.
European Journal of Organic Chemistry, 24, 5432-5437 (2015)
Synthesis of (S)-Tert-Butyl 3-Hydroxybutyrate by Asymmetric Reduction of Tert-Butyl Acetoacetate with Saccharomyces cerevisiae B5.
Ou ZM, et al.
Advanced Materials Research, 704 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service