Merck
  • Characterization of the commercially-available fluorescent chloroquine-BODIPY conjugate, LynxTag-CQGREEN, as a marker for chloroquine resistance and uptake in a 96-well plate assay.
Characterization of the commercially-available fluorescent chloroquine-BODIPY conjugate, LynxTag-CQGREEN, as a marker for chloroquine resistance and uptake in a 96-well plate assay.
PloS one (2014-10-25)
Cheryl C Y Loh, Rossarin Suwanarusk, Yan Quan Lee, Kitti W K Chan, Kit-Ying Choy, Laurent Rénia, Bruce Russell, Martin J Lear, François H Nosten, Kevin S W Tan, Larry M C Chow
ABSTRACT
Chloroquine was a cheap, extremely effective drug against Plasmodium falciparum until resistance arose. One approach to reversing resistance is the inhibition of chloroquine efflux from its site of action, the parasite digestive vacuole. Chloroquine accumulation studies have traditionally relied on radiolabelled chloroquine, which poses several challenges. There is a need for development of a safe and biologically relevant substitute. We report here a commercially-available green fluorescent chloroquine-BODIPY conjugate, LynxTag-CQGREEN, as a proxy for chloroquine accumulation. This compound localized to the digestive vacuole of the parasite as observed under confocal microscopy, and inhibited growth of chloroquine-sensitive strain 3D7 more extensively than in the resistant strains 7G8 and K1. Microplate reader measurements indicated suppression of LynxTag-CQGREEN efflux after pretreatment of parasites with known reversal agents. Microsomes carrying either sensitive- or resistant-type PfCRT were assayed for uptake; resistant-type PfCRT exhibited increased accumulation of LynxTag-CQGREEN, which was suppressed by pretreatment with known chemosensitizers. Eight laboratory strains and twelve clinical isolates were sequenced for PfCRT and Pgh1 haplotypes previously reported to contribute to drug resistance, and pfmdr1 copy number and chloroquine IC50s were determined. These data were compared with LynxTag-CQGREEN uptake/fluorescence by multiple linear regression to identify genetic correlates of uptake. Uptake of the compound correlated with the logIC50 of chloroquine and, more weakly, a mutation in Pgh1, F1226Y.
Sigma-Aldrich
Methiothepin mesylate salt, ≥98% (HPLC), solid
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
USP
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Desipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Mibefradil dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Sigma-Aldrich
Chlorprothixene hydrochloride
Desipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Chlorprothixene hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Desipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Loperamide hydrochloride, European Pharmacopoeia (EP) Reference Standard
Promethazine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Loperamide hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Promethazine hydrochloride
USP
Promethazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Loperamide hydrochloride
Supelco
Loperamide hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Octoclothepin maleate salt, solid
USP
Loperamide hydrochloride, United States Pharmacopeia (USP) Reference Standard