Skip to Content
Merck
All Photos(2)

Key Documents

C0982

Sigma-Aldrich

Chlorpromazine hydrochloride

meets USP testing specifications

Synonym(s):

2-Chloro-10-(3-dimethylaminopropyl)phenothiazine hydrochloride, CPZ, Largactil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2S · HCl
CAS Number:
Molecular Weight:
355.33
Beilstein:
3779989
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

98.0-101.5% dry basis

form

solid

mp

194-196  °C

solubility

water: soluble 50 g/L

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13

InChI

1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H

InChI key

FBSMERQALIEGJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.

Biochem/physiol Actions

Chlorpromazine demonstrates cytotoxic and antiproliferative activity against leukemic cells, but does not affect the viability of normal lymphocytes.
Phenothiazine antipsychotic; D2 dopamine receptor antagonist, H1 histamine receptor antagonist; inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endocytosis Is a Key Mode of Interaction between Extracellular I?-Amyloid and the Cell Membrane.
Shi, et al.
Biophysical Journal, 122, 1568-1569 (2023)
Xiaona Wei et al.
Veterinary research, 51(1), 10-10 (2020-02-12)
With the emergence of highly pathogenic variant strains, porcine epidemic diarrhea virus (PEDV) has led to significant economic loss in the global swine industry. Many studies have described how coronaviruses enter cells, but information on PEDV invasion strategies remains insufficient.
Rafaa Zeineddine et al.
Molecular neurodegeneration, 10, 57-57 (2015-11-02)
Amyotrophic Lateral Sclerosis is characterized by a focal onset of symptoms followed by a progressive spread of pathology that has been likened to transmission of infectious prions. Cell-to-cell transmission of SOD1 protein aggregates is dependent on fluid-phase endocytosis pathways, although
Mihail Lucian Pascu et al.
PloS one, 8(2), e55767-e55767 (2013-02-14)
Phenothiazines when exposed to white light or to UV radiation undergo a variety of reactions that result in degradation of parental compound and formation of new species. This process is slow and may be sped up with exposure to high
Rosanne Varkevisser et al.
Pflugers Archiv : European journal of physiology, 465(2), 247-259 (2012-11-30)
Drug-induced ion channel trafficking disturbance can cause cardiac arrhythmias. We showed that the antiprotozoic pentamidine decreased K(IR)2.x carried I(K1) current and that inhibiting protein degradation in the lysosome increased intracellular K(IR)2.1 levels. In this study, we aim to identify and

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service