Skip to Content
Merck
All Photos(4)

Key Documents

C3025

Sigma-Aldrich

(±)-Chlorpheniramine maleate salt

≥99% (perchloric acid titration)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (perchloric acid titration)

form

powder

mp

130-135 °C (lit.)

originator

Bayer

SMILES string

CN(C)CCC(C1=CC=C(Cl)C=C1)C2=NC=CC=C2.O=C(/C=C\C(O)=O)O

InChI

1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DBAKFASWICGISY-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Chlorpheniramine maleate (CPM)/(3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-yl-propan-1-amine is an alkylamine antihistamine.

Application

(±)-Chlorpheniramine maleate salt has been used:
  • as H1 receptor antagonist to determine the receptor function
  • to block the effect of compound 48/80 on plasma IGF-I
  • as a standard for fast sensing and determination by sequential injector coupled with potentiometer

Biochem/physiol Actions

Chlorpheniramine maleate (CPM) can prevent rhinitis and urticaria. It is used to cure several allergic conditions. This antihistamine is used in small-animal veterinary practices.
H1 Histamine receptor antagonist.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endogenous histamine reduces plasma insulin-like growth factor I via H1 receptor-mediated pathway in the rat
Liao W, et al.
European Journal of Pharmacology, 374(3), 471-476 (1999)
Sensing of chlorpheniramine in pharmaceutical applications by sequential injector coupled with potentiometer
Saleh TA
Journal of Pharmaceutical Analysis, 1(4), 246-250 (2011)
Functionalized Fe3O4 Magnetic Nanoparticle Potentiometric Detection Strategy versus Classical Potentiometric Strategy for Determination of Chlorpheniramine Maleate and Pseudoephedrine HCl
Moustafa AA, et al.
Journal of Analytical Methods in Chemistry (2019)
Extracellular signal-regulated kinase (ERK) activation is required for itch sensation in the spinal cord
Zhang L, et al.
Molecular Brain, 7(1), 25-25 (2014)
V Alagarsamy et al.
European journal of medicinal chemistry, 43(11), 2331-2337 (2007-11-23)
A series of novel 1-substituted-4-(2-methylphenyl)-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-ones were synthesized by the cyclization of 2-hydrazino-3-(2-methylphenyl)-3H-quinazolin-4-one with various one carbon donors. The starting material 2-hydrazino-3-(2-methylphenyl)-3H-quinazolin-4-one was synthesized from 2-methyl aniline by a novel innovative route. The title compounds were tested for their in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service