Merck
  • Occurrence and distribution of sulfonamides, tetracyclines, quinolones, macrolides, and nitrofurans in livestock manure and amended soils of Northern China.
Occurrence and distribution of sulfonamides, tetracyclines, quinolones, macrolides, and nitrofurans in livestock manure and amended soils of Northern China.
Environmental science and pollution research international (2014-10-17)
Jie Hou, Weining Wan, Daqing Mao, Chong Wang, Quanhua Mu, Songyan Qin, Yi Luo
ABSTRACT
A feasible and rapid analysis for the simultaneous determination of sulfonamides (SAs), tetracyclines (TCs), fluoroquinolones (FQs), macrolides (MACs) and nitrofurans (NFs) in livestock manure and soils was established by solid-phase extraction (SPE)-ultra-performance liquid chromatography (UPLC)-tandem mass spectrometry (MS/MS). A total of 32 manure and 17 amended soil samples from the Liaoning and Tianjin areas in Northern China were collected for analysis. The largest detected frequencies and concentrations in manure samples were those of TCs (3326.6 ± 12,302.6 μg/kg), followed by FQs (411.3 ± 1453.4 μg/kg), SAs (170.6 ± 1060.2 μg/kg), NFs (85.1 ± 158.1 μg/kg), and MACs (1.4 ± 4.8 μg/kg). In general, veterinary antibiotics (VAs) were detected with higher concentrations in swine and chicken manure than in cattle manure, reflecting the heavy usage of VAs in swine and chicken husbandry in the studied area. Furthermore, higher residues of antibiotics were found in piglet and fattening swine manure than in sow manure. In addition, TCs were the most frequently (100%) detected antibiotics in amended soil with higher concentrations (up to 10,967.1 μg/kg) than any other VAs. The attenuation of SAs was more obvious than TCs in amended soil after fertilization, which can most likely be attributed to the stronger sorption of TCs than SAs to soil organic matter through cation exchange. This study illustrated the prevalence of TCs detected in both animal manure and fertilized agricultural soils in Northern China, which may increase the risk to human health through the food chain. Thus, TCs should be given more attention in the management of veterinary usage in livestock husbandry.
Product Description
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Sigma-Aldrich
Sulfamonomethoxine
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Supelco
Sulfamonomethoxine, VETRANAL®, analytical standard
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Furazolidone
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Supelco
Sulfamethoxazole
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sulfadimidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sulfadiazine for identification of impurity F, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadimidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadiazine, European Pharmacopoeia (EP) Reference Standard
Sulfamethoxazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfadiazine, Pharmaceutical Secondary Standard; Certified Reference Material
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
USP
Sulfadiazine, United States Pharmacopeia (USP) Reference Standard
USP
Sulfamethoxazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard