F9505
Furazolidone
Synonym(s):
3-(5-Nitrofurfurylideneamino)-2-oxazolidinone
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C8H7N3O5
CAS Number:
Molecular Weight:
225.16
Beilstein:
8317414
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Recommended Products
biological source
synthetic
form
powder
color
yellow
mp
256-256 °C
solubility
formic acid: 50 mg/mL
antibiotic activity spectrum
Gram-positive bacteria
parasites
Mode of action
enzyme | inhibits
SMILES string
[O-][N+](=O)c1ccc(\C=N\N2CCOC2=O)o1
InChI
1S/C8H7N3O5/c12-8-10(3-4-15-8)9-5-6-1-2-7(16-6)11(13)14/h1-2,5H,3-4H2/b9-5+
InChI key
PLHJDBGFXBMTGZ-WEVVVXLNSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Furazolidone is a nitrofuran derivative with antiprotozoal and antibacterial activity. Furazolidone binds bacterial DNA, which leads to the gradual inhibition of monoamine oxidase. It is used to treat anorexia and antagonism of thiamin utilization in poultry. Furazolidone increases thapsigargin-sensitive Ca2+-ATPase in chick cardiac myocytes.
Biochem/physiol Actions
Furazolidone and its generated free radicals, may bind to DNA and induce cross-links. Bacterial DNA is particularly susceptible to this drug, which results in high levels of mutations (transitions and transversions) in the bacterial chromosome. Its mechanism of action minimizes the development of resistant organisms. Furazolidone is a monoamine oxidase (MAO) inhibitor and is used as a DNA interstrand cross-linking agent.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
B H Ali et al.
Quarterly journal of experimental physiology (Cambridge, England), 67(3), 437-448 (1982-07-01)
Furazolidone (0.4% w/w in the feed, 10 days) reduced the feed intake and growth in 9 week old chickens, and increased the amount of 5-hydroxytryptamine (5-HT) in the brain. The drug also increased the stimulation of transketolase activity by thiamine
Am J Physiol Heart Circ Physiol. Furazolidone increases thapsigargin-sensitive Ca2+-ATPase in chick cardiac myocytes
D. Lax, R. Martinez-Zaguilan, et al.
American Journal of Physiology. Heart and Circulatory Physiology, 267, H734-H741 (1994)
B H Ali
Veterinary research communications, 6(1), 1-11 (1983-01-01)
The pharmacological and toxicological properties of furazolidone have been briefly reviewed. Among the most important pharmacological actions of furazolidone is the inhibition of mono- and diamine oxidase activities, which seem to depend, at least in some species, on the presence
In vivo and in vitro metabolic studies of furazolidone: a risk evaluation.
L H Vroomen et al.
Drug metabolism reviews, 22(6-8), 663-676 (1990-01-01)
André Gustavo Tempone et al.
International journal of antimicrobial agents, 36(2), 159-163 (2010-06-18)
Drug delivery systems are promising pharmaceutical formulations used to improve the therapeutic index of drugs. In this study, we developed a liposomal formulation of furazolidone that targets Leishmania (Leishmania) chagasi amastigotes in a hamster model. Using laser scanning confocal microscopy
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service