Skip to Content
Merck
All Photos(1)

Key Documents

1478108

USP

Ofloxacin

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ofloxacin, ofloxacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C(=CN1c24)C(O)=O

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)

InChI key

GSDSWSVVBLHKDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ofloxacin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Ofloxacin Tablets
  • Ofloxacin Ophthalmic Solution
  • Levofloxacin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahesh D Chavanpatil et al.
International journal of pharmaceutics, 316(1-2), 86-92 (2006-03-29)
Oral sustained release gastroretentive dosage forms offer many advantages for drugs having absorption from upper gastrointestinal tract and improve the bioavailability of medications that are characterized by a narrow absorption window. A new gastroretentive sustained release delivery system was developed
J P Monk et al.
Drugs, 33(4), 346-391 (1987-04-01)
Ofloxacin is one of a new generation of fluorinated quinolones structurally related to nalidixic acid. It is an orally administered broad spectrum antibacterial drug active against most Gram-negative bacteria, many Gram-positive bacteria and some anaerobes. Ciprofloxacin is the only other
G A Gates
The Pediatric infectious disease journal, 20(1), 104-107 (2001-02-15)
To assess the safety of topical agents in the middle ear, animal studies were reviewed. Compared with aminoglycoside-containing preparations, which caused significant loss of hair cells in the basal turn of the cochlea, ofloxacin caused no loss of hair cells
Shu-Hwa Hsiao et al.
The Annals of pharmacotherapy, 39(1), 146-149 (2004-11-25)
To report a case of ofloxacin/levofloxacin-induced rhabdomyolysis and to compare other reported cases from the literature. A 19-year-old male patient developed ofloxacin/levofloxacin-induced rhabdomyolysis during admission for periorbital cellulitis. Symptoms of myalgia, weakness, and swelling of the arms developed after 3
R J Ruben
The Pediatric infectious disease journal, 20(1), 108-110 (2001-02-15)
Ofloxacin otic is as effective in curing otitis externa in children (<12 years of age) as is the preparation combining polymyxin/neomycin plus hydrocortisone and as ciprofloxacin otic. Furthermore ofloxacin otic, as it requires less frequent dosing, is likely to encourage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service