Merck
  • Enhanced signal intensity in matrix-free laser desorption ionization mass spectrometry by chemical modification of bionanostructures from diatom cell walls.
Enhanced signal intensity in matrix-free laser desorption ionization mass spectrometry by chemical modification of bionanostructures from diatom cell walls.
Rapid communications in mass spectrometry : RCM (2014-05-28)
Tino Jaschinski, Kathleen Thume, Moritz Klein, Phillipp Richter, Jürgen Popp, Aleš Svatoš, Georg Pohnert
ABSTRACT
Laser desorption ionization for mass spectrometric measurements (LDI MS) is supported by nanostructured materials. This technique helps to overcome known limitations of matrix-assisted laser desorption/ionization (MALDI) and especially avoids interfering signals caused by matrix components. LDI can be supported by bionanostructures from the cell walls of diatoms. We explore how ionization efficiency can be improved by chemical modification of the cell walls. We introduce procedures to chemically modify these nanopatterned silicate structures using perfluorooctyldimethylchlorosilane or pentafluorophenylpropyldimethylchlorosilane. Using a conventional MALDI-MS instrument we compare ionization using the novel materials with that of unmodified cell walls. The functionalized bionanomaterial is comprehensively evaluated for the use in LDI MS using a broad range of analytes and two commercial drugs. Chemical modifications lead to materials that support LDI significantly better than unmodified diatom cell walls. LDI signal intensity was up to 25-fold increased using the modified preparations. No interfering signals in the lower molecular weight range down to m/z 100 were observed, demonstrating the suitability of the method for small analytes. Crude solutions of commercial drugs, such as Aspirin complex(®) and IbuHEXAL(®) could be directly investigated without additional sample preparation. Chemically modified diatom cell walls represent a powerful tool to support ionization in LDI MS. The lack of background signals in the low molecular weight region of the mass spectra allows also the investigations of small analytes.
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, for chromatography
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%