Skip to Content
Merck
All Photos(1)

Key Documents

110086

Sigma-Aldrich

Acetonitrile

99%, suitable for UV/Vis spectroscopy, ReagentPlus®

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12191502
eCl@ss:
39031501
PubChem Substance ID:
NACRES:
NA.03
grade:
reagent
technique(s):
UV/Vis spectroscopy: suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

Product Name

Acetonitrile, ReagentPlus®, 99%

grade

reagent

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

UV/Vis spectroscopy: suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

organic solvents: soluble(lit.)
water: soluble(lit.)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile (MeCN), an aliphatic nitrile, is a colorless liquid with a characteristic odor. MeCN has low viscosity, high elution strength and is highly miscible in water. It is widely used as a solvent and intermediate in organic syntheses and is transparent to UV-visible light, making it highly applicable in spectrophotometric and fluorimetric techniques. It also plays a major role as an extractant medium in liquid-liquid extraction, solid-phase extraction or microextraction.

Application


  • Metal-cation-induced shifts in thiolate redox and reduced sulfur speciation.: This study explores the redox behavior of thiolates affected by metal cations in acetonitrile, shedding light on sulfur speciation processes critical for developing catalytic and environmental remediation technologies (Seo et al., 2024).

  • Rational design and investigation of nonlinear optical response properties of pyrrolopyrrole aza-BODIPY-based novel push-pull chromophores.: Focused on nonlinear optics, this research uses acetonitrile as a solvent to study the optical properties of newly designed chromophores, contributing to advances in optical materials science (Kothoori et al., 2024).

  • Acetonitrile real gas phase behavior from quasi-ideal gas to nanodroplets: A microscopical view.: Providing a microscopic insight into the phase behavior of acetonitrile, this research enhances understanding of fluid dynamics at nanoscale levels, relevant for both academic studies and industrial applications (Martínez et al., 2024).

  • Crystalline CdS/amorphous Cd(OH)2 Composite for Electrochemical CO2 Reduction to CO in a Wide Potential Window.: Highlighting the role of acetonitrile in electrochemical setups, this study develops new materials for efficient CO2 reduction, potentially impacting energy conversion strategies globally (Hua et al., 2024).


Features and Benefits

ReagentPlus® solvents have a guaranteed purity ≥98.5% and are ideal for experiments and applications that need fewer specifications. They are primarily defined by assay specifications and offer suitable quality for general lab use.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 6-6 (1994)
Purification of acetonitrile.
Walter M and Ramaley L.
Analytical Chemistry, 45(1), 165-166 (1973)
Timo Glatter et al.
Molecular systems biology, 5, 237-237 (2009-01-22)
Protein complexes represent major functional units for the execution of biological processes. Systematic affinity purification coupled with mass spectrometry (AP-MS) yielded a wealth of information on the compendium of protein complexes expressed in Saccharomyces cerevisiae. However, global AP-MS analysis of
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations
Divya Subramonian et al.
Journal of proteome research, 13(9), 3905-3918 (2014-07-30)
SUMOylation is an essential posttranslational modification and regulates many cellular processes. Dysregulation of SUMOylation plays a critical role in metastasis, yet how its perturbation affects this lethal process of cancer is not well understood. We found that SUMO-2/3 modification is

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service