Merck
  • Unsaturated long-chain fatty acids inhibit the binding of oxidized low-density lipoproteins to a model CD36.
Unsaturated long-chain fatty acids inhibit the binding of oxidized low-density lipoproteins to a model CD36.
Bioscience, biotechnology, and biochemistry (2014-07-19)
Marie Takai, Yuki Kozai, Satoshi Tsuzuki, Yukari Matsuno, Maiko Fujioka, Kozue Kamei, Hitomi Inagaki, Ai Eguchi, Shigenobu Matsumura, Kazuo Inoue, Tohru Fushiki
ABSTRACT
Transmembrane protein CD36 binds multiple ligands, including oxidized low-density lipoproteins (oxLDLs) and long-chain fatty acids (LCFAs). Our aim was to determine whether LCFAs compete with oxLDLs for binding to CD36. We addressed this issue by examining the inhibitory effect of LCFAs against the binding of Alexa-fluor-labeled oxLDLs (AFL-oxLDL) to a synthetic peptide representing the oxLDL-binding site on CD36 (3S-CD36₁₅₀₋₁₆₈). All of the unsaturated LCFAs tested, inhibited the binding of AFL-oxLDL to 3S-CD36₁₅₀₋₁₆₈, albeit to varying degrees. For instance, the concentrations required for 50% inhibition of binding for oleic, linoleic, and α-linolenic acids were 0.25, 0.97, and 1.2 mM, respectively. None of the saturated LCFAs tested (e.g. stearic acid) exhibited inhibitory effects. These results suggest that at least unsaturated LCFAs can compete with oxLDLs for binding to CD36. The study also provides information on the structural requirements of LCFAs for inhibition of oxLDLs-CD36 binding.
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Methyl oleate, technical grade, 70%
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Methyl oleate, 99%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Supelco
Methyl oleate, analytical standard
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, analytical standard
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Supelco
Oleic acid, Selectophore, ≥99%
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Oleic acid, European Pharmacopoeia (EP) Reference Standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Oleyl alcohol, technical grade, 85%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Oleyl alcohol, ≥99% (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%