Skip to Content
Merck
All Photos(4)

Key Documents

311111

Sigma-Aldrich

Methyl oleate

99%

Synonym(s):

Methyl cis-9-octadecenoate, Oleic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CO2CH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727037
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10 mmHg ( 205 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

218 °C/20 mmHg (lit.)

density

0.874 g/mL at 20 °C (lit.)

functional group

ester

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10-

InChI key

QYDYPVFESGNLHU-KHPPLWFESA-N

Looking for similar products? Visit Product Comparison Guide

General description

The methyl oleate monolayers at the air-water interface undergoes ozonolysis that results in rapid loss of material through cleavage of the C=C bond and evaporation/dissolution of reaction products.

Application

Methyl oleate can be used as a reactant to synthesize:
  • Epoxidized methyl oleate via epoxidation.
  • Allyl azides via manganese(III) acetate-mediated addition of azide in the alkyl chain.
  • Bromohydrin products via hydroxybromination with N-bromosuccinimide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Dei Cas et al.
Frontiers in pharmacology, 11, 570616-570616 (2020-12-29)
Cannabis (Cannabis sativa L.) is a highly promising medicinal plant with well-documented effectiveness and growing use in the treatment of various medical conditions. Cannabis oils are mostly used in galenic preparations, due to their easy adjustment of the administration dose
Estefania Oliva et al.
Biomolecules, 10(2) (2020-02-26)
Bearing grafts based on fatty esters derivatives, lipidyl-cyclodextrins (L-CDs) are compounds able to form water-soluble nano-objects. In this context, bicatenary biobased lipidic-cyclodextrins of low DS were easily synthesized from a fatty ester epoxide by means of alternative methods (ball-milling conditions
Tatiana R Amarante et al.
Inorganic chemistry, 52(8), 4618-4628 (2013-03-28)
The reaction of MoO3, 2,2'-bipyridine-5,5-dicarboxylic acid (H2bpdc), water, and dimethylformamide in the mole ratio 1:1:1730:130 at 150 °C for 3 days in a rotating Teflon-lined digestion bomb leads to the isolation of the molybdenum oxide/bipyridinedicarboxylate hybrid material (DMA)[MoO3(Hbpdc)]·nH2O (1) (DMA
Wan-Ying Wong et al.
The Science of the total environment, 727, 138534-138534 (2020-04-26)
Interest in biodiesel research has escalated over the years due to dwindling fossil fuel reserves. The implementation of a carbon-based solid acid catalyst in biodiesel production eradicates the separation problems associated with homogeneous catalysis. However, its application in the glycerol-free
Aman Ullah et al.
ChemSusChem, 10(10), 2167-2174 (2017-03-02)
Catalytic transformation of renewable feedstocks into fine chemicals is in high demands and olefin metathesis is a sophisticated tool for biomass conversion. Nevertheless, the large-scale viability of such processes depends on the conversion efficiency, energy efficiency, catalytic activity, selective conversion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service