Skip to Content
Merck
All Photos(1)

Key Documents

1621008

USP

Stearic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

1 mmHg ( 173.7 °C)

API family

stearic acid

manufacturer/tradename

USP

bp

361 °C (lit.)

mp

67-72 °C (lit.)

solubility

acetone: soluble
benzene: freely soluble
chloroform: freely soluble
ethanol: soluble
ether: freely soluble
propylene glycol: soluble
water: practically insoluble

density

0.845 g/cm3

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Stearic acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Palmitic Acid
  • Calcium Stearate
  • Sodium Stearate
  • Aluminum Monostearate
  • Glycerin Suppositories
  • Magnesium Stearate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of USP Biologics Program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aluminum Monostearate
United States Pharmacopeia, 44(4), 5612-5612 (2019)
Stearic acid
United States Pharmacopeia, 34(5), 3146-3146 (2020)
Palmitic Acid
United States Pharmacopeia, 42(3), 5918-5918 (2019)
Sodium Stearate
United States Pharmacopeia, 39(3), 6024-6024 (2023)
S M Grundy
The American journal of clinical nutrition, 60(6 Suppl), 986S-990S (1994-12-01)
Stearic acid is a long-chain saturated fatty acid. However, in contrast with other saturated fatty acids, stearic acid apparently does not raise serum cholesterol concentrations. Studies carried out three decades ago provided strong suggestive evidence that this was the case.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service