Merck
  • Infrared biospectroscopy for a fast qualitative evaluation of sample preparation in metabolomics.
Infrared biospectroscopy for a fast qualitative evaluation of sample preparation in metabolomics.
Talanta (2014-06-11)
Julia Kuligowski, David Pérez-Guaita, Javier Escobar, Isabel Lliso, Miguel de la Guardia, Bernhard Lendl, Máximo Vento, Guillermo Quintás
ABSTRACT
Liquid chromatography-mass spectrometry (LC-MS) has been increasingly used in biomedicine to study the dynamic metabolomic responses of biological systems under different physiological or pathological conditions. To obtain an integrated snapshot of the system, metabolomic methods in biomedicine typically analyze biofluids (e.g. plasma) that require clean-up before being injected into LC-MS systems. However, high resolution LC-MS is costly in terms of resources required for sample and data analysis and care must be taken to prevent chemical (e.g. ion suppression) or statistical artifacts. Because of that, the effect of sample preparation on the metabolomic profile during metabolomic method development is often overlooked. This work combines an Attenuated Total Reflectance-Fourier transform infrared (ATR-FTIR) and a multivariate exploratory data analysis for a cost-effective qualitative evaluation of major changes in sample composition during sample preparation. ATR-FTIR and LC-time of flight mass spectrometry (TOFMS) data from the analysis of a set of plasma samples precipitated using acetonitrile, methanol and acetone performed in parallel were used as a model example. Biochemical information obtained from the analysis of the ATR-FTIR and LC-TOFMS data was thoroughly compared to evaluate the strengths and shortcomings of FTIR biospectroscopy for assessing sample preparation in metabolomics studies. Results obtained show the feasibility of ATR-FTIR for the evaluation of major trends in the plasma composition changes among different sample pretreatments, providing information in terms of e.g., amino acids, proteins, lipids and carbohydrates overall contents comparable to those found by LC-TOFMS.
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%