Merck
  • Fibulin-2 is involved in early extracellular matrix development of the outgrowing mouse mammary epithelium.
Fibulin-2 is involved in early extracellular matrix development of the outgrowing mouse mammary epithelium.
Cellular and molecular life sciences : CMLS (2014-02-14)
D Olijnyk, A M Ibrahim, R K Ferrier, T Tsuda, M-L Chu, B A Gusterson, T Stein, J S Morris
ABSTRACT
Cell-matrix interactions control outgrowth of mammary epithelium during puberty and pregnancy. We demonstrate here that the glycoprotein fibulin-2 (FBLN2) is strongly associated with pubertal and early pregnant mouse mammary epithelial outgrowth. FBLN2 was specifically localized to the cap cells of the terminal end buds during puberty and to myoepithelial cells during very early pregnancy (days 2-3) even before morphological changes to the epithelium become microscopically visible, but was down-regulated thereafter. Exposure to exogenous oestrogen (E2) or E2 plus progesterone (P) increased Fbln2 mRNA expression in the pubertal gland, indicating hormonal control. FBLN2 was co-expressed and co-localised with the proteoglycan versican (VCAN) and co-localised with laminin (LN), while over-expression of FBLN2 in HC-11 cells increased cell adhesion to several extracellular matrix proteins including LN and fibronectin, but not collagens. Mammary glands from Fbln2 knockout mice showed no obvious phenotype but increased fibulin-1 (FBLN1) staining was detected, suggesting a compensatory mechanism by other fibulin family members. We hypothesise that similar to embryonic aortic smooth muscle development, FBLN2 and VCAN expression alters the cell-matrix interaction to allow mammary ductal outgrowth and development during puberty and to enable epithelial budding during pregnancy.
Product Description
SAFC
BIS-TRIS
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-FBLN2 antibody produced in rabbit
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
DAPI, for nucleic acid staining