Merck
  • Nonextractable residue formation of sulfonamide antimicrobials: new insights from soil incubation experiments.
Nonextractable residue formation of sulfonamide antimicrobials: new insights from soil incubation experiments.
Chemosphere (2014-01-28)
Anna Gulkowska, Basilius Thalmann, Juliane Hollender, Martin Krauss
ABSTRACT
Soil incubation experiments using (14)C-labelled sulfamethazine were carried out to assess the factors governing its nonextractable residue (NER) formation via nucleophilic addition reactions. Circumstantial evidence on possible mechanisms of NER formation was derived from a selective manipulation of soil samples. The amount of quinones in soil available for nucleophilic addition was a limiting factor as indicated by (i) an (initial) increase of NER formation by adding quinone precursors or enhancing their formation by manganese oxide addition and (ii) a decrease of NER formation by limiting the formation of quinones under anaerobic conditions. A slow NER formation with time under aerobic conditions is likely caused by covalent bonding as well, because no slow NER formation phase was observed under anaerobic conditions.
Product Description
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydroquinone, JIS special grade, ≥99.0%
Sigma-Aldrich
Manganese(II,III) oxide, 97%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sulfadimidine for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Sulfadimidine, European Pharmacopoeia (EP) Reference Standard