Skip to Content
Merck
All Photos(1)

Documents

13-1930

Sigma-Aldrich

Hydroquinone

JIS special grade, ≥99.0%

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

JIS special grade

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

Assay

≥99.0%

form

crystalline

autoignition temp.

930 °F

availability

available only in Japan

pH

3.7 (70 g/L)

bp

285 °C (lit.)

mp

172-175 °C (lit.)

solubility

water: soluble 50 g/L

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob Levitt
Journal of the American Academy of Dermatology, 57(5), 854-872 (2007-05-01)
Recently, the US Food and Drug Administration proposed a ban on over-the-counter hydroquinone mainly on the basis of high absorption, reports of exogenous ochronosis in humans, and murine hepatic adenomas, renal adenomas, and leukemia with large doses over extended time
Yaqi Hu et al.
Biosensors & bioelectronics, 47, 45-49 (2013-04-03)
A novel light "on-off" photoelectrochemical sensing for the sensitive determination of hydroquinone was developed based on the porphyrin-functionalized Au nanoparticles on graphene (porphyrin/AuNPs/graphene). Gold nanoparticles (AuNPs) were firstly modified onto graphene sheets using NaBH4 as reductant and the porphyrin/AuNPs/graphene nanocomposites
P Carbonnelle et al.
Toxicology and applied pharmacology, 132(2), 220-226 (1995-06-01)
Decreased interleukin-1 (IL-1) production by mononuclear phagocytes has been shown to contribute to benzene myelotoxicity in animals. The study presented here was designed to examine the relevance of this mechanism in humans. Fresh human blood monocytes were exposed to 0.001-10
Nuan P Cheah et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(3), 1072-1081 (2013-02-19)
Tobacco smoke consists of thousands of harmful components. A major class of chemicals found in tobacco smoke is formed by aldehydes, in particular formaldehyde, acetaldehyde and acrolein. The present study investigates the gene expression changes in human lung alveolar epithelial
Douglas McGregor
Critical reviews in toxicology, 37(10), 887-914 (2007-11-21)
The toxicology of hydroquinone has been reviewed on a number of previous occasions. This review targets its potential for carcinogenicity and possible modes of carcinogenic action. The evaluation made by IARC (1999) of its carcinogenic risk to humans was that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service