Merck
The use of glycolic acid as a peeling agent.
Dermatologic clinics (1995-04-01)
H Murad, A T Shamban, P S Premo
ABSTRACT
Glycolic acid is a member of the AHA family, which occurs naturally in foods and has been used for centuries as a cutaneous rejuvenation treatment. Recently it has proved to be a versatile peeling agent and it is now widely used to treat many defects of the epidermis and papillary dermis in a variety of strengths, ranging from 20% to 70%, depending on the condition being treated. People of almost any skin type and color are candidates, and almost any area of the body can be peeled. Several weeks prior to a peel the skin may be prepared with topical tretinoin or glycolic acid, and immediately prior to the peel the skin may be degreased with a variety of agents. Following the peel the skin is carefully observed for any complications such as hyperpigmentation and infection. Results are maintained with serial peels and at-home use of tretinoin or glycolic acid, as well as sun avoidance. The glycolic acid can be applied simultaneously with TCA and is another technique for a medium-depth peel. Comparison of 35% TCA-treated skin with 70% glycolic acid-treated skin examined histologically at different times reveals similar changes in papillary dermis connective tissue proteins, epidermal necrosis seen only with TCA, and reversion at 2 years postpeel to pretreatment appearance.
Product Description
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Supelco
Glycolic Acid, Pharmaceutical Secondary Standard; Certified Reference Material