Skip to Content
Merck
All Photos(1)

Key Documents

12-1300

Sigma-Aldrich

Glycolic acid solution

CP, 70% in H2O

Synonym(s):

Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COOH
CAS Number:
Molecular Weight:
76.05
Beilstein:
1209322
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

CP

form

liquid

availability

available only in Japan

concentration

70% in H2O

SMILES string

OCC(O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W H Porter et al.
Journal of toxicology. Clinical toxicology, 39(6), 607-615 (2002-01-05)
To correlate serum glycolic acid levels with clinical severity and outcome in ethylene glycol poisoning and to determine if glycolic acid levels are predictive of renal failure and the need for hemodialysis. We measured serum ethylene glycol and glycolic acid
Thea R Pick et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(8), 3185-3190 (2013-02-06)
Photorespiratory carbon flux reaches up to a third of photosynthetic flux, thus contributes massively to the global carbon cycle. The pathway recycles glycolate-2-phosphate, the most abundant byproduct of RubisCO reactions. This oxygenation reaction of RubisCO and subsequent photorespiration significantly limit
Ingela Bjurhager et al.
Biomacromolecules, 13(8), 2521-2527 (2012-07-10)
In 1628, the Swedish warship Vasa capsized on her maiden voyage and sank in the Stockholm harbor. The ship was recovered in 1961 and, after polyethylene glycol (PEG) impregnation, it was displayed in the Vasa museum. Chemical investigations of the
D Jacobsen et al.
The American journal of medicine, 84(1), 145-152 (1988-01-01)
Ethylene glycol and glycolate kinetics were studied in two cases of ethylene glycol intoxication with maximal ethylene glycol/glycolate concentrations of 40.9/26.8 and 56.4/22.4 mmol/liter, respectively. Both patients survived, but with prolonged renal failure, upon treatment with bicarbonate, ethanol, and hemodialysis.
Genqiang Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 24(34), 4609-4613 (2012-06-26)
Novel ZnMn(2)O(4) ball-in-ball hollow microspheres are fabricated by a facile two-step method involving the solution synthesis of ZnMn-glycolate hollow microspheres and subsequent thermal annealing in air. When evaluated as an anode material for lithium-ion batteries, these ZnMn(2)O(4) ball-in-ball hollow microspheres

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service