Skip to Content
Merck
All Photos(1)

Documents

12-1290

Sigma-Aldrich

Glycolic acid

SAJ special grade, ≥98.0%

Synonym(s):

Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COOH
CAS Number:
Molecular Weight:
76.05
Beilstein:
1209322
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

form

crystalline

availability

available only in Japan

mp

75-80 °C (lit.)

SMILES string

OCC(O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>572.0 °F - (decomposition)

Flash Point(C)

> 300 °C - (decomposition)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thea R Pick et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(8), 3185-3190 (2013-02-06)
Photorespiratory carbon flux reaches up to a third of photosynthetic flux, thus contributes massively to the global carbon cycle. The pathway recycles glycolate-2-phosphate, the most abundant byproduct of RubisCO reactions. This oxygenation reaction of RubisCO and subsequent photorespiration significantly limit
W H Porter et al.
Journal of toxicology. Clinical toxicology, 39(6), 607-615 (2002-01-05)
To correlate serum glycolic acid levels with clinical severity and outcome in ethylene glycol poisoning and to determine if glycolic acid levels are predictive of renal failure and the need for hemodialysis. We measured serum ethylene glycol and glycolic acid
Ingela Bjurhager et al.
Biomacromolecules, 13(8), 2521-2527 (2012-07-10)
In 1628, the Swedish warship Vasa capsized on her maiden voyage and sank in the Stockholm harbor. The ship was recovered in 1961 and, after polyethylene glycol (PEG) impregnation, it was displayed in the Vasa museum. Chemical investigations of the
Genqiang Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 24(34), 4609-4613 (2012-06-26)
Novel ZnMn(2)O(4) ball-in-ball hollow microspheres are fabricated by a facile two-step method involving the solution synthesis of ZnMn-glycolate hollow microspheres and subsequent thermal annealing in air. When evaluated as an anode material for lithium-ion batteries, these ZnMn(2)O(4) ball-in-ball hollow microspheres
D Jacobsen et al.
The American journal of medicine, 84(1), 145-152 (1988-01-01)
Ethylene glycol and glycolate kinetics were studied in two cases of ethylene glycol intoxication with maximal ethylene glycol/glycolate concentrations of 40.9/26.8 and 56.4/22.4 mmol/liter, respectively. Both patients survived, but with prolonged renal failure, upon treatment with bicarbonate, ethanol, and hemodialysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service