Merck
Methanol and formic acid toxicity: biochemical mechanisms.
Pharmacology & toxicology (1991-09-01)
J Liesivuori, H Savolainen
ABSTRACT
Metabolism of methanol, methyl ethers, esters and amides give rise to formic acid. This acid is an inhibitor of the mitochondrial cytochrome oxidase causing histotoxic hypoxia. Formic acid is a weaker inhibitor than cyanide and hydrosulphide anions. The body burden of formate in methanol poisoning is high enough to cause acidosis, and other clinical symptoms. Part of the protons can be attributed to formic acid whereas the most significant acid load results from the hypoxic metabolism. The acidosis causes e.g. dilatation of cerebral vessels, facilitation of the entry of calcium ions into cells, loss of lysosomal latency and deranged production of ATP. The latter effect seems to impede parathormone-dependent calcium reabsorption in the kidney tubules. Besides, urinary acidification is affected by formic acid. Its excretion causes continuous recycling of the acid by the tubular cell Cl-/formate exchanger. This sequence of events may partially explain an accumulation of formate in urine. Occupational exposure to vapours of methanol and formic acid can be quantitatively monitored by urinary formic acid determinations. Formic acid toxicity may prove a suitable model for agents causing histotoxic hypoxia.
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%