Merck
  • Review on toxicity testing with marine macroalgae and the need for method standardization--exemplified with copper and phenol.
Review on toxicity testing with marine macroalgae and the need for method standardization--exemplified with copper and phenol.
Marine pollution bulletin (2003-02-15)
Britta T Eklund, Lena Kautsky
ABSTRACT
Toxic effects on macroalgae have been compiled. Eighty-two articles have been found in literature during 1959-2000. A total of 120 substances were investigated using 65 different macroalgae species. About one-third of the tested compounds were organic substances (33%), another third metal-organic substances (35%), and the last third were oils (14%), metals (8%), detergents (7.5%) and other inorganic chemicals (2.5%). Half of the substances were only tested once on a single species. Likewise, toxicity data has only been reported for one chemical tested on a single occasion for about half of the 65 species. Thus little is known about the toxic effects on marine macroalgae. The objectives of the previous studies undertaken varied and therefore the toxicity data was presented in numerous ways, e.g. using different exposure times, temperature, light intensity, light regime, salinity, and nutrients, which makes a direct comparison of the data difficult. This review also shows that many stages in the lifecycle of macroalgae are often more sensitive to toxic substances than other aquatic organisms. Consequently, tests using macroalgae may discover toxicity earlier, which would in turn also protect the fauna. If toxic compounds have a negative affect on the distribution and growth of structurally and functionally dominating macroalgae, there may indirectly be a large and harmful influence on the whole marine coastal ecosystem. For this reason tests on macroalgae should be included in control programs along the coasts.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Pandy′s reagent
Sigma-Aldrich
Phenol stock solution, 100 mg/dL, standard