Merck
  • Synthesis, structure and gas-sensing properties of Pd-functionalized ZnSnO3 rods.
Synthesis, structure and gas-sensing properties of Pd-functionalized ZnSnO3 rods.
Journal of nanoscience and nanotechnology (2013-05-08)
Hyunsu Kim, Changhyun Jin, Sunghoon Park, Chongmu Lee
ABSTRACT
ZnSnO3 one-dimensional (1D) strutures were synthesized by using an evaporation technique. The morphology, crystal structure, and enhanced sensing properties of the ZnSnO3 structures functionalized with Pd to CO gas at 300 degrees C were investigated. The diameters of the 1D structures ranged from a few hundreds to a few thousands of nanometers and that the lengths were up to a few hundreds of micrometers. The gas sensors fabricated from multiple networked ZnSnO3 rods functionalized with Pd showed the enhanced electrical responses to CO gas. The responses of the rods were improved 10.7, 13.7, 13.4, and 12.5 fold at the CO concentrations of 10, 25, 50, and 100 ppm, respectively. In addition, the mechanism for the enhancement in the gas sensing properties of ZnSnO3 rods by Pd functionalization is discussed.
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis