Skip to Content
Merck
All Photos(3)

Key Documents

203998

Sigma-Aldrich

Palladium

powder or granules, 99.99% trace metals basis

Synonym(s):

Palladium (powder), Palladium black, Palladium element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder or granules

resistivity

9.96 μΩ-cm, 20°C

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palladium(Pd) is a silver-white noble transition metal in the platinum group of metals.Relative to the platinum group metals, Pd has the lowest density, 12 g/cm3, andlowest melting point, 1555 °C. Pd is known for its strong catalytic activityand is used in various applications such as electrical equipment, dentalmaterials, implanted medical devices, and automobile catalysts. Additionally,it is also extensively employed in the preparation of diverse heterogeneouspalladium catalysts for various organic transformations. Our high-purity Pdpowder is differentiated by its high trace metals purity, which makes itparticularly useful in sensitive research projects in electronics, catalysis,and synthesis.

Application

  • The Effect P Additive on the CeZrAl Support Properties and the Activity of the Pd Catalysts in Propane Oxidation.: This study explores the role of phosphorus additives on the properties of CeZrAl supports and their impact on palladium catalyst activity in propane oxidation. Findings suggest that phosphorus improves the structural stability and enhances the oxidation activity of the palladium catalysts, making it a significant factor in energy storage solutions and industrial applications (Feng et al., 2024).
  • A Multifunctional Co-Based Metal-Organic Framework as a Platform for Proton Conduction and Nitrophenols Reduction.: While primarily focusing on cobalt-based frameworks, this paper discusses palladium′s role as a comparative catalyst. It highlights palladium′s efficiency in proton conduction and nitrophenol reduction, indicating its potential for energy-efficient industrial processes and renewable energy applications (Ma et al., 2023).
  • Two-Dimensional Lanthanide Metal-Organic Frameworks as a Platform for Sensing Pollutant and Nitrophenols Reduction.: Investigates the application of palladium within two-dimensional lanthanide metal-organic frameworks for pollutant sensing and nitrophenol reduction, highlighting its utility in environmental monitoring and energy conversion systems (Wang et al., 2023).
  • Core-Shell, Critical-Temperature-Suppressed V Alloy-Pd Alloy Hydrides for Hydrogen Storage-A Technical Evaluation.: Evaluates the use of vanadium alloy-palladium alloy hydrides in hydrogen storage, focusing on their technical properties and potential for enhancing energy storage efficiency. This study provides insights into developing high-purity palladium-based materials for energy conversion and storage (Lamb et al., 2023).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work
Stefano Nicolai et al.
The Journal of organic chemistry, 78(8), 3783-3801 (2013-03-21)
Tetrahydrofurans and pyrrolidines are among the most important heterocycles found in bioactive compounds. Cyclization-functionalization domino reactions of alcohols or amines onto olefins constitute one of the most efficient methods to access them. In this context, oxy- and aminoalkynylation are especially
Xuxing Chen et al.
Organic & biomolecular chemistry, 11(16), 2582-2585 (2013-03-19)
A one-pot two-step synthesis of versatile indenones has been developed. This palladium(II)-catalyzed transformation involves generation and condensation of ortho-functionalized 1,2-benzils from 2-(2-arylethynylphenyl)acetonitriles using Ph2SO as the oxidant. The resulting 3-cyanoindenones can be converted to various valuable molecules.
Meifang Zheng et al.
Organic letters, 15(8), 1838-1841 (2013-04-02)
Palladium-catalyzed oxidative difunctionalization of enol ethers with 1,3-dicarbonyl compounds to construct trisubstituted furans in one step under mild conditions is described. The reaction is thought to proceed through a C-C bond formation along with a C-O bond closing the ring
Meng Wang et al.
Organic & biomolecular chemistry, 11(16), 2574-2577 (2013-03-15)
A novel Pd/Cu catalyzed domino reaction for the synthesis of functionalized pyrrolo[1,2-b]pyridazines from readily accessible (hetero)aryl propargyl alcohols and 1-amino-2-bromopyrroles was developed. This cascade process involves a Sonogashira cross-coupling reaction, an isomerization and an intramolecular condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service